Oxoid™ Oxford Listeria Selective Supplement
Actual product may vary
Thermo Scientific™

Oxoid™ Oxford Listeria Selective Supplement

The Oxoid Listeria Selective Supplement (Oxford) is used for the detection of Listeria monocytogenes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberYield
SR0206BFor 2.5L medium
SR0206EEach vial for 500mL medium
Catalog number SR0206B
Price (INR)
-
Yield:
For 2.5L medium

Oxoid Listeria Selective Supplement (Oxford) is used for the detection of Listeria monocytogenes.

  • Add to Listeria Selective Agar Base, Part No. CM0856B
Specifications
DescriptionMod Lister(oxf)sup For 2.5L 10Pk.
YieldFor 2.5L medium
Product TypeSelective Supplement
SpeciesListeria
Unit SizeEach