Oxoid™ Modified Brucella Selective Supplement
Thermo Scientific™
Oxoid™ Modified Brucella Selective Supplement
Oxoid Modified Brucella Selective Supplement is for the cultivation and isolation of Brucella spp.
Have Questions?
Catalog NumberQuantity
SR0209E10/Pk.
Catalog number SR0209E
Price (INR)
-
Quantity:
10/Pk.
Oxoid Modified Brucella Selective Supplement is for the cultivation and isolation of Brucella spp.
  • Alternative supplement containing natamycin as a replacement for cycloheximide
  • Add to Blood Agar Base No. 2, Part No. CM0271B or Brucella Medium Base, Part No. CM0169B
  • Each vial supplements 500mL of medium
For Laboratory Use
For Laboratory Use
Specifications
DescriptionModified Brucella Selective Supplement
Quantity10/Pk.
YieldEach vial for 500mL medium
FormFreeze-dried
Product TypeSelective Supplement
SpeciesBrucella species
Unit SizeEach