Product Image
Thermo Scientific™
Oxoid™ Modified Listeria Selective Enrichment Supplement
The Oxoid Listeria Selective Supplement, Modified (Modified Listeria Selective Supplement) is an alternative supplement containing amphotericin B as a replacement for cycloheximide.
Have Questions?
Catalog NumberQuantity
SR0213E10/Pk.
Catalog number SR0213E
Price (INR)
-
Quantity:
10/Pk.
Oxoid Listeria Selective Supplement, (Modified Listeria Selective Supplement) is an alternative supplement containing amphotericin B as a replacement for cycloheximide.
Add to to Buffered Listeria Enrichment Broth, Part No. CM0897R or Listeria Enrichment Broth, Part No. CM0862B. For the enrichment of Listeria monocytogenes. Each vial supplements 500mL of medium.
Specifications
DescriptionListeria Selective Supplement, Modified
Quantity10/Pk.
YieldEach vial for 500mL medium
FormFreeze-dried
Product TypeSelective Supplement
SpeciesListeria
Unit SizeEach