Oxoid™ Campylobacter Growth Supplement
Thermo Scientific™
Oxoid™ Campylobacter Growth Supplement
Oxoid Campylobacter Growth Supplement is a liquid supplement for the enhanced growth and aerotolerance of Campylobacter species.
Have Questions?
Catalog NumberQuantity
SR0232E10/Pk.
Catalog number SR0232E
Price (INR)
-
Quantity:
10/Pk.
Oxoid Campylobacter Growth Supplement is a liquid supplement for the enhanced growth and aerotolerance of Campylobacter species.
  • For the preparation of Blaser-Wang Medium, Bolton Selective Enrichment Broth, Butzler Selective Medium, and Preston Selective Medium
  • Each vial supplements 500mL of medium
Specifications
DescriptionCampylobacter Growth Supplement
Quantity10/Pk.
YieldEach vial for 500mL medium
FormFreeze-dried
Product TypeMicrobiology Media
SpeciesCampylobacter
Unit SizeEach