Oxoid™ ONE Broth-Listeria Supplement
Thermo Scientific™

Oxoid™ ONE Broth-Listeria Supplement

The Oxoid ONE Broth-Listeria is for the selective enrichment broth for Listeria species from food samples in 24 hours.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberYield
SR0234BEach vial for 2.25L medium
SR0234HEach vial for 2 L medium
SR0234EEach vial for 500mL medium
Catalog number SR0234B
Price (INR)
-
Yield:
Each vial for 2.25L medium

The Oxoid ONE Broth-Listeria is for the selective enrichment broth for Listeria species from food samples in 24 hours.

Add to ONE Broth-Listeria, Part No. CM1066.

ONE Broth - Listeria: Typical Formulation

  • Peptone: 26g/L; Carbohydrate mix: 6g/L; Salt mix: 10g/L with a final pH of 7.4 ±0.2 at 25°C

Appearance:

  • Dehydrated medium: straw coloured, free-flowing powder
  • Selective supplement: orange/yellow, freeze-dried pellet
  • Prepared medium: dark straw coloured, clear liquid

Note:

Formulation Directions

  • Suspend 22g of ONE Broth-Listeria Base in 500mL (44g/L) of distilled water
  • Mix well and sterilize by autoclaving at 121°C for 15 minutes
  • Cool the medium to below 50°C and aseptically add the contents of one vial of ONE Broth-Listeria Selective Supplement (Mfr. No. SR0234) reconstituted as directed

Testing Protocol

  • Add 25g of food sample to 225mL of ONE Broth-Listeria and stomach for a minimum of 30 seconds to mix the sample
  • Incubate the broth without agitation at 30°C for 24±2 hours
  • Gently agitate the bag then, using a microbiological loop, remove 10μL and inoculate onto a suitable agar medium e.g. Oxoid Brilliance Listeria Agar (ISO) (Mfr. No. CM1212, SR257 and SR0258) and incubate appropriately (37°C for 24-48 hours)

  • Confirm presumptive colonies on the agar plates as Listeria monocytogenes or Listeria spp. by appropriate methods e.g. Gram-stain, catalase, oxidase, Oxoid Listeria Latex Test Kit (Mfr. No. DR1126)
  • Oxoid O.B.I.S. mono (Mfr. No. ID0600), Microbact Listeria 12L (Mfr. No. MB1128)

 


For Laboratory Use Only

Formulation Directions

  • Suspend 22g of ONE Broth-Listeria Base in 500mL (44g/L) of distilled water
  • Mix well and sterilize by autoclaving at 121°C for 15 minutes
  • Cool the medium to below 50°C and aseptically add the contents of one vial of ONE Broth-Listeria Selective Supplement (Mfr. No. SR0234) reconstituted as directed

Testing Protocol

  • Add 25g of food sample to 225mL of ONE Broth-Listeria and stomach for a minimum of 30 seconds to mix the sample
  • Incubate the broth without agitation at 30°C for 24±2 hours
  • Gently agitate the bag then, using a microbiological loop, remove 10μL and inoculate onto a suitable agar medium e.g. Brilliance Listeria Agar (Mfr. No. CM1080, SR0222, and SR0228) and incubate appropriately (37°C for 24 to 48 hours)
  • Confirm presumptive colonies on the agar plates as Listeria monocytogenes or Listeria spp. by appropriate methods e.g. Gram-stain, catalase, oxidase, Oxoid Listeria Latex Test Kit DR1126, Oxoid O.B.I.S. mono ID0600, Microbact Listeria 12L MB1128

General References:

  1. ISO 11290-1 (Microbiology of food and animal feeding stuffs — Horizontal method for the enumeration of Listeria monocytogenes — part 1).
  2. Oxoid Poster - Folio No. 1033, July 2004.
  3. Data on file at Oxoid
Specifications
DescriptionONE Broth-Listeria Supplement
Quantity10/Pk.
YieldEach vial for 2.25L medium
FormFreeze-dried
Product TypeMicrobiology Media
SpeciesListeria
Unit SizeEach