Search
Search
| Catalog Number | Type | Electrical Requirements | Includes |
|---|---|---|---|
| SRF110P1-230 | Vacuum Concentrator Kit | 230 V, 50 Hz, 8 A | Oil Pump, Vapor Trap, Accessories |
| RF108 | 12-13 mm Rotor | N/A | Rotor |
| RF132 | Hexagonal Microcentrifuge Rotor | N/A | Rotor |
| RF32 | 28 mm Rotor | N/A | Rotor |
| RFMT4 | Microplate Rotor | N/A | Rotor |
| SRF110-115 | Vacuum Concentrator | 115 V, 60 Hz, 16 A | Concentrator |
| SRF110-230 | Vacuum Concentrator | 230 V, 50 Hz, 8 A | Concentrator |
| SRF110P1-115 | Vacuum Concentrator Kit | 115 V, 60 Hz, 16 A | Oil Pump, Vapor Trap, Accessories |
| SRF110P2-115 | Vacuum Concentrator Universal Vacuum Kit | 115 V, 60 Hz, 16 A | Universal Vacuum System, Rotor, Accessories |
| SRF110P2-230 | Vacuum Concentrator Universal Vacuum Kit | 230 V, 50 Hz, 8 A | Universal Vacuum System, Rotor, Accessories |
SpeedVac™ SRF110 modular refrigerated vacuum concentrator provides flexibility to your lab. It offers unique programming capabilities to maintain chamber temperature pre and post sample run to prevent thermal degradation of heat sensitive samples such as proteins, enzymes, and other biomolecules. They are ideal for evaporation of aqueous based solvents, buffers, and HPLC fractions. For a convenient, one model shopping experience select from SRF110P1 (modular configuration) and SRF110P2 (Semi integrated configuration) kit options. Each kit is designed to include components required for convenient SpeedVac setup.
SRF110 modular refrigerated vacuum concentrator provides flexibility to any benchtop space. SRF110 can be configured depending on your applications need. Select from wide range of suitable Thermo Scientific components such as pumps, Vapor Traps, and additional accessories to configure your custom vacuum concentration setup. For a convenient, one model shopping experience select from SRF110P1 (modular configuration) and SRF110P2 (Semi integrated configuration) kit options. Each kit is designed to include components required for convenient SpeedVac setup.