2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 3 ports, Luer Lock insert and body connectors, injection site
Thermo Scientific™

2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 3 ports, Luer Lock insert and body connectors, injection site

The 1 L Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, aRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
SS00005I
also known as SS00005-I
2 L
SS00004I
also known as SS00004-I
1 L
Catalog number SS00005I
also known as SS00005-I
Price (INR)
-
Capacity (Metric):
2 L
The 1 L Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL, animal origin–free ethyl vinyl acetate film and includes 3 ports (1 insert Luer Lock + cap, 1 body Luer Lock + cap, 1 injection site).

Features of the system include:
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications include:
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquid
• Collecting chromatography feed and fraction
• Harvesting, storing, and transporting bulk drug product and drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, and steam-to and steam-through connectors
• Filters (used for bioburden reduction)—sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers and platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands

Specifications
Dimensions33.3 cm x 20.3 cm (13.1 in. x 8.0 in.)
Outer PackagingOne 4-mil. poly bag
Product LineLabtainer
Quantity2 L, 50 per case
RegulatorycGMP
Shipping ConditionRoom Temperature
Sterilization MethodIrradiated
Capacity (Metric)2 L
Film DescriptionASI 28
MaterialEVA
Port DetailLuer Lock Body Connector and Injection Site
Tubing StyleEVA
Unit Size2 L
Contents & Storage
Store at room temperature.