2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, Luer Lock insert and body connectors with reducers, injection site
Thermo Scientific™

2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, Luer Lock insert and body connectors with reducers, injection site

The Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layerRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
SS00033I
also known as SS00033-I
2 L
SS00029I
also known as SS00029-I
100 mL
SS00030I
also known as SS00030-I
250 mL
SS00031I
also known as SS00031-I
500 mL
SS00032I
also known as SS00032-I
1 L
Catalog number SS00033I
also known as SS00033-I
Price (INR)
-
Capacity (Metric):
2 L

The Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL polyethylene film.

Features
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• Constructed in an ISO 7 clean room under cGMP conditions

Applications
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquids
• Collecting chromatography feeds and fractions
• Harvesting, storing, and transporting bulk drug product and drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters (used for bioburden reduction )—sterilization grade filters, vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style ports, needle-free ports
• Clamps—sliders, pinch clamps, crimp bands

BPC port specifications:
Line 1: Luer lock insert connection with 4.8 mm (3/16 in.) barb, cap, slide clamp, and 4.8 x 4.1 mm (3/16 x 5/32 in.) reducer. Tubing: TPE 374, length: 30 cm (12 in.), ID x OD: 3.2 x 6.4 mm (0.13 x 0.25 in.)
Line 2: Luer lock body connection with 0.56 cm (0.22 in.) barb, plug, slide clamp, and 0.48 x 0.41 cm (0.12 x 0.16 in.) reducer. Tubing: TPE 374, length: 30 cm (12 in.), ID x OD: 3.2 x 6.4 mm (0.13 x 0.25 in.)
Line 3: Tubing: EVA, injection port

Specifications
Dimensions30.5 x 20.3 cm (12.0 x 8.0 in.)
Outer PackagingOne 4-mil. poly bag
Product LineLabtainer
Product TypeBioProcess Container
Quantity50/Cs.
Shipping ConditionRoom Temperature
Sterilization MethodIrradiated
Capacity (Metric)2 L
Film DescriptionASI 26/77
MaterialPolyethylene
Port Detail3 Ports (1 Luer Lock Insert Connection + Cap, 1 Luer Lock Body Connection + Cap, 1 Injection Site)
Tubing StyleTPE and EVA
Unit Size2 L
Contents & Storage
Store at room temperature.