2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, MPC insert and body connectors, injection site
Thermo Scientific™

2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, MPC insert and body connectors, injection site

The 100 mL Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, aRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
SS00037I
also known as SS00037-I
1 L
SS00034I
also known as SS00034-I
100 mL
SS00035I
also known as SS00035-I
250 mL
SS00036I
also known as SS00036-I
500 mL
SS00038I
also known as SS00038-I
2 L
Catalog number SS00037I
also known as SS00037-I
Price (INR)
-
Capacity (Metric):
1 L
The 100 mL Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL polyethylene film. This BPC includes 3 ports (1 MPC insert + cap, 1 MPC body + cap, 1 injection site).

Features
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• Constructed in an ISO 7 clean room under cGMP conditions

Applications
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquids
• Collecting chromatography feeds and fractions
• Harvesting, storing, and transporting bulk drug product and drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters (used for bioburden reduction )—sterilization grade filters, vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style ports, needle-free ports
• Clamps—sliders, pinch clamps, crimp bands

BPC port specifications:
• Line 1: MPC insert with 6.4 mm (1/4 in.) barb, cap, and slide clamp. Tubing: EVA + TPE 374
• Line 2: MPC body with 6.4 mm (1/4 in.) barb, plug, and slide clamp. Tubing: EVA + TPE 374
• Line 3: Tubing: EVA, injection port

Specifications
Dimensions32 cm x 14.7 cm (12.6 in. x 5.8 in.)
Outer Packaging4 mil. poly bag, One 4-mil. poly bag
Product LineLabtainer
Quantity30 per case
Shipping ConditionRoom Temperature
Sterilization MethodIrradiated
Capacity (Metric)1 L
Film DescriptionASI 26/77
MaterialPolyethylene
Port DetailMPC Connector and Injection Port
Tubing StyleTPE and EVA
Unit Size1 L
Contents & Storage
Store at room temperature.