2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 3 ports, MPC insert connectors and break-off injection site
Thermo Scientific™

2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 3 ports, MPC insert connectors and break-off injection site

The 100 mL Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, aRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
SS00068I
also known as SS00068-I
500 mL
SS00066I
also known as SS00066-I
100 mL
SS00067I
also known as SS00067-I
250 mL
SS00069I
also known as SS00069-I
1 L
SS00070I
also known as SS00070-I
2 L
Catalog number SS00068I
also known as SS00068-I
Price (INR)
-
Capacity (Metric):
500 mL
The 100 mL Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL, animal origin–free ethyl vinyl acetate film and includes 3 ports (2 MPC insert + cap, 1 break-off injection site).

Features of the system include:
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications include:
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquid
• Collecting chromatography feed and fraction
• Harvesting, storing, and transporting bulk drug product and drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, and steam-to and steam-through connectors
• Filters (used for bioburden reduction)—sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers and platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands

Specifications
Dimensions20.3 cm x 16.5 cm (8.0 in. x 6.5 in.)
Outer PackagingOne 4-mil. poly bag
Product LineLabtainer
Quantity50 per case
RegulatorycGMP
Shipping ConditionRoom Temperature
Sterilization MethodIrradiated
Capacity (Metric)500 mL
Film DescriptionASI 28
MaterialEVA
Port DetailMPC insert and body connectors and injection site
Tubing StyleEVA
Unit Size500 mL
Contents & Storage
Store at room temperature.