2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 2 ports, MPX insert and ReadyMate connector
Thermo Scientific™

2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 2 ports, MPX insert and ReadyMate connector

The 50 L Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, aRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
SS00155I
also known as SS00155-I
50 L
SS00156I
also known as SS00156-I
100 L
SS00157I
also known as SS00157-I
200 L
Catalog number SS00155I
also known as SS00155-I
Price (INR)
-
Capacity (Metric):
50 L
The 50 L Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL film. This BPC includes 2 ports (1 MPX insert + cap, 1 ReadyMate DAC).

Features
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• Constructed in an ISO 7 clean room under cGMP conditions

Applications
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquids
• Collecting chromatography feeds and fractions
• Harvesting, storing, and transporting bulk drug product and drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters (used for bioburden reduction )—sterilization grade filters, vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style ports, needle-free ports
• Clamps—sliders, pinch clamps, crimp bands

BPC port specifications:
• Line 1: MPX insert with 12.7 mm (1/2 in.) barb + cap, pinch clamp. Tubing: Silicone, length: 51 cm (20 in.), ID x OD: 12.7 x 19.1 mm (0.5 x 0.75 in.)
• Line 2: ReadyMate DAC with 12.7 mm (1/2 in.) barb, pinch clamp. Tubing: Silicone, length: 91.4 cm (36 in.), ID x OD: 12.7 x 19.1 mm (0.5 x 0.75 in.)

Specifications
Dimensions95.3 cm x 54.6 cm (37.5 in. x 21.5 in.)
For Use With (Equipment)TK-0050-05
Outer Packaging4 mil. poly bag, One 4-mil. poly bag
Product LineLabtainer
Protective Inner PackagingPoly bags on tubing ends, protective material around rough areas
Quantity10 per case
Shipping ConditionRoom Temperature
Sterilization MethodIrradiated
Capacity (Metric)50 L
Film DescriptionASI 26/77
MaterialPolyethylene
Port DetailMPC insert + cap, ReadyMate DAC connections
Tubing StyleSilicone
Unit Size50 L
Contents & Storage
Store at room temperature.