2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, capsule filter, MPC insert, injection port
Thermo Scientific™

2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, capsule filter, MPC insert, injection port

The 20 L Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, aRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
SS00160I
also known as SS00160-I
10 L
SS00158I
also known as SS00158-I
1 L
SS00159I
also known as SS00159-I
5 L
SS00161I
also known as SS00161-I
20 L
Catalog number SS00160I
also known as SS00160-I
Price (INR)
-
Capacity (Metric):
10 L
The 20 L Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL film. This BPC includes 3 ports (1 membrane capsule filter, MPC insert, pinch clamps, and an injection site).

Features
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• Constructed in an ISO 7 clean room under cGMP conditions

Applications
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquids
• Collecting chromatography feeds and fractions
• Harvesting, storing, and transporting bulk drug product and drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters (used for bioburden reduction )—sterilization grade filters, vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style ports, needle-free ports
• Clamps—sliders, pinch clamps, crimp bands

BPC port specifications:
• Line 1: 0.2 μm PES membrane capsule filter + pinch clamp. Tubing: TPE 374, length: 30.5 cm (12 in.), ID x OD: 9.5 x 15.9 mm (0.38 x 0.63 in.)
• Line 2: MPC insert + cap, pinch clamp. Tubing: TPE 374, length: 91.4 cm (36 in.), ID x OD: 9.5 x 15.9 mm (0.38 x 0.63 in.)
• Line 3: Injection site

Specifications
Capacity (Metric)10 L
Dimensions (L x W x H)32 x 56.5 cm (12.6 x 22.3 in.)
Film DescriptionASI™ 26/77
Includes3 Ports (1 Luer Lock Insert Connection + Cap, 1 Luer Lock Body Connection + Cap, 1 Injection Site)
Length (Metric)32 cm
Outer PackagingTwo 4-mil. poly bags
Port DetailCapsule filter and MPC insert + cap
Product LineLabtainer
Protective Inner PackagingProtective material around rough areas
Quantity15/Cs.
Shipping ConditionRoom Temperature
Sterilization MethodIrradiated
Tubing StyleTPE
Width (Metric)56.5 cm
MaterialPolyethylene
Product TypeBioprocess Container
Width (English)22.3 in.
Unit Size10 L
Contents & Storage
Store at room temperature.