Product Image
Thermo Scientific™
Stainless Steel Interior Option for Thermo Scientific™ ULT Freezers
Choose a stainless steel interior option for your freezer with an optional, factory-installed stainless steel interior.
Have Questions?
Catalog NumberFor Use With (Equipment)
SSPERFORMANCEThermo Scientific ULT Freezers
Catalog number SSPERFORMANCE
Price (INR)
-
For Use With (Equipment):
Thermo Scientific ULT Freezers
Choose a stainless steel, factory-installed interior option for your Thermo Scientific™ Revco™ ExF, Forma™ 900 Series, HERAfreeze™ HFU-B or TSE Series -86°C upright freezers.
  • Factory installed
  • Stainless steel interior
† Factory-installed, specify at time of freezer order
Specifications
DescriptionStainless-steel interior
For Use With (Equipment)Thermo Scientific ULT Freezers
TypeUltra Low Freezer Interior , Stainless Steel
StyleUpright
Temperature Range-86°C
Unit SizeEach