NuPAGE™ Midi Bis-Tris Welcome Pack, 4 to 12%
Invitrogen™

NuPAGE™ Midi Bis-Tris Welcome Pack, 4 to 12%

The NuPAGE Bis-Tris Midi Welcome Pack provides all of the necessary NuPAGE gels, buffers, and reagents needed to begin using the SureLock Tandem Midi Gel Tank.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWells
STM400226-well
STM400312 + 2-well
STM400120-well
Catalog number STM4002
Price (INR)
67,291.00
Precio exclusivo compra web
Ends: 30-Sep-2026
122,346.61
Save 55,055.61 (45%)
1 pack
Wells:
26-well
Price (INR)
67,291.00
Precio exclusivo compra web
Ends: 30-Sep-2026
122,346.61
Save 55,055.61 (45%)
1 pack
The NuPAGE Bis-Tris Midi Welcome Pack provides all of the necessary NuPAGE gels, buffers, and reagents needed to begin using the SureLock Tandem Midi Gel Tank. The SureLock Tandem Midi Gel Tank enables rapid electrophoresis of midi gels using minimal buffer (∼520 mL/gel) in a leak-free system. With a setup time of ∼30 seconds, the tank efficiently runs midi gels while also providing consistent performance. The tank features two independent chambers, allowing electrophoresis of one or two gels at a time, which saves on buffer and limits waste.

The NuPAGE Bis-Tris Midi Welcome Pack contains:

  • SureLock Tandem Midi Gel Tank (STM1001)
  • NuPAGE Bis-Tris Midi Gels (2 boxes, 20 gels)
  • NuPAGE MES SDS Running Buffer, 20X (NP0002)
  • MES and MOPS Running Buffer, Powder (NP000105, NP000205)
  • NuPAGE LDS Sample Buffer, 4X (NP0007)
  • NuPAGE Sample Reducing Agent, 10X (NP0009)
  • PageRuler Plus Prestained Protein Ladder, 10 to 250 kDa (26619)

About the SureLock Tandem Midi Gel Tank

The SureLock Tandem Midi Gel Tank enables rapid electrophoresis of midi gels using minimal buffer (∼520 mL/gel) in a leak-free system. With a setup time of ∼30 seconds, the tank efficiently runs midi gels while also providing consistent performance. The tank features two independent chambers, allowing electrophoresis of one or two gels at a time, which saves on buffer and limits waste. The tank is compatible with all Invitrogen pre-cast midi gels and midi gel cassettes.

About NuPAGE Bis-Tris Midi Gels

NuPAGE Bis-Tris Midi protein gels are precast polyacrylamide gels designed to give optimal separation of a wide range of proteins under denaturing conditions. Unlike traditional Tris-glycine midi gels, NuPAGE Bis-Tris midi gels have a neutral pH environment that minimizes protein modifications. Use NuPAGE Bis-Tris Midi gels for preparing proteins for sequencing, mass spectrometry, and any other techniques where protein integrity is crucial. Also, use NuPAGE midi gels for optimal results during day-to-day use.

Transfer and detection

For transfer of proteins to a membrane, we recommend using NuPAGE Transfer Buffer (NP0006) for traditional wet transfer using the SureLock Tandem Midi Blot Module (STM2001). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)SureLock Tandem Midi Gel Tank
Gel Thickness1.0 mm
Quantity1 Welcome Pk.
Shipping ConditionApproved for shipment at room temperature and wet ice
Gel Percentage4 to 12%
Gel SizeMidi
Gel TypeBis-Tris
Product LineNuPAGE
Wells26-well
Unit Size1 pack