DynaSpin™ Single-Use Rotor, harvest
Thermo Scientific™
DynaSpin™ Single-Use Rotor, harvest
This DynaSpin consumable is for use with the DynaSpin Single-Use Centrifuge. It is composed of a rotor, a tube setRead more
Have Questions?
Catalog NumberQuantity
SUT000561 each
Catalog number SUT00056
Price (INR)
-
Quantity:
1 each
This DynaSpin consumable is for use with the DynaSpin Single-Use Centrifuge. It is composed of a rotor, a tube set that implements disk stack technology, and three line sets with integrated best-in-class sensors that allow analysis and control of critical process parameters. It has one inlet line where cell culture fluid is supplied to the rotor and two outlet lines that carry the separated product (centrate) and concentrate streams. Each line set contains appropriate sensors that communicate with the hardware via the firmware on the built-in touchscreen.
Specifications
AutoclavableNot Autoclavable
For Use With (Equipment)DynaSpin™ Single-Use Centrifuge
Quantity1 each
Unit Size1 each