HyPerforma™ Mixtainer™ Hardware, Conical Drum, 100L
Thermo Scientific™

HyPerforma™ Mixtainer™ Hardware, Conical Drum, 100L

The 100L Thermo Scientific™ Conical Drum, top dispense, with clamps is utilized as part of the Thermo Scientific™ HyPerforma™ Mixtainer™ system to support bioproduction mixing applications.
Have Questions?
Catalog NumberQuantity
SV50517.12100 L
Catalog number SV50517.12
Price (INR)
-
Quantity:
100 L

The 100L Thermo Scientific™ Conical Drum, top dispense, with clamps is utilized as part of the Thermo Scientific™ HyPerforma™ Mixtainer™ system to support bioproduction mixing applications. Additional components of the system include the Thermo Scientific™ HyPerforma™ Mixtainer™ BioProcess Container (BPC) with an integrated magnetic stir bar, a magnetic stirring plate, a support plate and a dolly. The optimized support hardware allows mixing, moving, storage, shipping and discharge of the BPC contents while maintaining sterility of the contents. Standard products are available for sterile liquid-to-liquid or powder-to-liquid mixing.

Recommended for:

  • Hydration of powdered media and buffers
  • Sterile mixing of cell culture media with sera and reagents
  • Sterile mixing of buffer solutions or other process fluids
  • Pooling of sterile liquid fractions
Specifications
Dimensions60 cm x 76 cm (23.5 in. x 29.75 in.)
Port DetailBottom Port: 10.2 cm (4 in.)
Product LineHyPerforma, Mixtainer
Quantity100 L
Capacity (Metric)100 L
Unit Size100 L