HyPerforma™ Mixtainer™ Hardware, Conical Drum, 50L
HyPerforma™ Mixtainer™ Hardware, Conical Drum, 50L
Thermo Scientific™

HyPerforma™ Mixtainer™ Hardware, Conical Drum, 50L

The 50 L Thermo Scientific™ Conical Drum, top dispense, with clamps is utilized as part of the Thermo Scientific™ HyPerforma™Read more
Have Questions?
Catalog NumberQuantity
SV505171150 L
Catalog number SV5051711
Price (INR)
-
Request A Quote
Quantity:
50 L

The 50 L Thermo Scientific™ Conical Drum, top dispense, with clamps is utilized as part of the Thermo Scientific™ HyPerforma™ Mixtainer™ system to support bioproduction mixing applications. Additional components of the system include the Thermo Scientific™ HyPerforma™ Mixtainer™ BioProcess Container (BPC) with an integrated magnetic stir bar, a magnetic stirring plate, a support plate and a dolly. The optimized support hardware allows mixing, moving, storage, shipping and discharge of the BPC contents while maintaining sterility of the contents. Standard products are available for sterile liquid-to-liquid or powder-to-liquid mixing.

Recommended for:
• Hydration of powdered media and buffers
• Sterile mixing of cell culture media with sera and reagents
• Sterile mixing of buffer solutions or other process fluids
• Pooling of sterile liquid fractions

Specifications
Dimensions23.5 x 23 in. (60 x 58 cm)
Port DetailBottom Port: 4 in. (10.2 cm)
Product LineHyPerforma, Mixtainer
Quantity50 L
Capacity (Metric)50 L
Unit Size50 L