Smart-Vue Pro Receivers
Thermo Scientific™
Smart-Vue Pro Receivers
IT friendly LoRaWan provides easy set-up of a reliable, independent and secure wireless network
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
SVPHWAPG868Smart-Vue Pro MTC PRO Ethernet/4G LoRa Gateway Receiver with Antenna, Power Supply, 865-868 Mhz
SVPHWAPG868CSmart-Vue Pro MTC PRO Ethernet/4G LoRa Gateway Receiver with SIM card, data plan for up to 10 devices/1 year. Includes Antenna, Power Supply, 865-868 Mhz
SVPHWAPG915Smart-Vue Pro MTC PRO Ethernet/4G LoRa Gateway Receiver with Antenna, Power Supply, 915 Mhz, no data/SIM
SVPHWAPG915CSmart-Vue Pro MTC PRO Ethernet/4G LoRa Gateway Receiver with SIM card, data plan for up to 10 devices/1 year. Includes Antenna, Power Supply, 915 Mhz
Catalog number SVPHWAPG868
Price (INR)
-
Includes:
Smart-Vue Pro MTC PRO Ethernet/4G LoRa Gateway Receiver with Antenna, Power Supply, 865-868 Mhz
LoRA is short for Long Range wireless Wide Area Network. It is a stand-alone dedicated network and increases connectivity range for IoT devices by 2 km in urban and 10 km in rural areas. This receiver pushes data to the Cloud by connecting to your local network via an Ethernet or Wi-Fi connection.
Long range wireless connectivity enables a single receiver to cover large areas, replacing the need for repeaters Dedicated wireless network requires only an internet connection, minimal IT support needed to set up. Support for up to 1000 data logging sensors per gateway
Specifications
CalibrationNA
Data MemoryNA
For Use With (Equipment)Smart-Vue Pro Remote Monitoring
IncludesSmart-Vue Pro MTC PRO Ethernet/4G LoRa Gateway Receiver with Antenna, Power Supply, 865-868 Mhz
ModelPRO MTC Model for up to 50 Modules
Quantity1
Warranty1 Year Warranty
Alarm TypesE-mail, SMS, Audible, Telephone, Mobile Push Notification
Display TypeNo Display
TypeRemote Monitoring Gateway
Unit SizeEach