InSight Software
Thermo Scientific™
InSight Software
Thermo Scientific™ InSightLink Premisis Manager works with your WLAN and the InSight Cloud.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
SW-INSIGHT-1002InSightLink Premisis Manager, server software (WLAN -> InSight Cloud). (Additional site with Master)
SW-RENEW-1005Yr >=2 Annual subscription renewal, ultra-low freezer
SW-INSIGHT-1001InSightLink Premisis Manager, server software (WLAN -> InSight Cloud). Master Site
SW-RENEW-1006
Catalog number SW-INSIGHT-1002
Price (INR)
-
Description:
InSightLink Premisis Manager, server software (WLAN -> InSight Cloud). (Additional site with Master)
Thermo Scientific™ InSight software tools work with Insight motes and sensors to monitor your cold storage equipment’s temperature energy consumption, and other parameters using Wi-Fi or proprietary wireless protocols.
  • Warranty varies by country, please contact us for details
Not available outside North America
Specifications
DescriptionInSightLink Premisis Manager, server software (WLAN -> InSight Cloud). (Additional site with Master)
TypeInsight Software
Unit SizeEach