Product Image
Thermo Scientific™

Area Radiation Monitoring System for Nuclear Power Plants

The turnkey, drop-and-go Thermo Scientific™ Area Monitoring Package for NPP is ideal for situations where simple and rapid area monitoring setup is needed.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
SYS171110002
also known as SYS/1/71110/002
Area Monitoring Package
SYS171110001
also known as SYS/1/71110/001
Area Monitoring Package with ViewPoint
Catalog number SYS171110002
also known as SYS/1/71110/002
Price (INR)
-
Description:
Area Monitoring Package

The turnkey, drop-and-go Thermo Scientific™ Area Monitoring Package for Nuclear Power Plants for nuclear power facilities is ideal for situations where simple and rapid area monitoring setup is needed. The package is designed for rapid deployment for remote monitoring use in a wired (LAN) or wireless environment.

Thermo Scientific Area Radiation Monitoring System incorporates the highly versatile and functional Thermo Scientific CNET1500 Network Docking Station Gamma Monitor (CNET 1500 NDS) with the industry leading software monitoring system—Thermo Scientific ViewPoint Enterprise™.

Components and Key Features of Package:

CNET1500 NDS (with EPD)

  • Wired (LAN) or wireless (2.4 GHz) connectivity to ViewPoint server
  • Point-to-point or radio base station mode (for connecting to other CNETs or EPD transmitters)
  • Multi-range EPD sensor
  • Configurable alarm set points
  • Audible and visual alarms
  • Drop-and-go setup

ViewPoint™ Enterprise Software System

  • ViewPoint server and single client workstation
  • EPD + CNET1500 NDS Plug-ins
  • Fully networkable configuration - remote viewing capable
  • Email/text interface
  • Full graphing and historical data analysis

CNET1500 W Wireless Network Base

  • 2.4 GHz wireless base capability
  • LAN compatible
  • Remote antenna capable

EPD-G Gamma Radiation Detector

  • 0.5 mR/h to 30 R/h range
  • Internal alarms/self diagnostics

Onsite support/training

  • Complimentary ViewPoint training class (half-day)
  • Configuration and Field Set Up Assistance (half-day)

Includes:

  • CNET1500 NDS (with EPD)
  • ViewPoint Enterprise Software System
  • CNET1500 W Wireless Network Base
  • EPG-G Gamma Radiation Dosimeter
  • Onsite support/training

Recommended for:

  • Radiation waste handling activities
  • Outage related activities: Steam generator platform coverage; Fuel handling activities; Reactor maintenance activities such as CRDM maintenance, reactor incore/seal table, reactor cavity, etc.; Monitoring of high rad areas to streamline and improve HRA entry control
Specifications
ApprovalsUS FCC: Part 15.247; EU: ETS 300.328; License free
Audible AlarmMode: Ultra Loud Continuous Tone; Sound Pressure: 108 dBA at 15 VDC, 24 inches;
Backup Battery9.6 Volt Ni-MH, 8xSC, 3000 mAh
Cabinet MaterialWatertight, injection molded polycarbonate
Channel Data Rate460 Kbps
ChannelsUSA: 75; France: 33; Spain: 25; Japan: 25
CurrentPower Supply: 100 mA; Strobe Light: 150 mA at 12 VDC; Sound Alarm: 30 mA at 5 VDC
DescriptionArea Monitoring Package
Error Correction24-bit CRC and ARQ
Frequency Band Details2400 to 2483 MHz (USA); 2448 to 2480 MHz (France)
I/O Data RateUp to 230 Kbps
IncludesCNET1500 NDS (with EPD); ViewPoint Enterprise Software System; CNET1500 W Wireless Network Base; EPG-G Gamma Radiation Dosimeter; Onsite support/training
Interface BoardRadio Interface; RS-232 Asynchronous
MaterialWatertight, injection molded polycarbonate
Modulation TypeGMSK
Network ProtocolARQ: Dynamically assigned TDMA
Network TopologyStar
Operating Range (English)Indoor: 450 to 900 ft.; Outdoor w/ Supplied Antenna: 3000 ft. (w/ Gain Antenna: >6 miles)
Operating Range (Metric)Indoor: 137 m to 275 m; Outdoor: 915 m with supplied
Output Impedance50Ω
ProcessorCNET 1500
Receiver Sensitivity-93 dBm for 10-5 BER
Sensitivity0.5 mR/h-30R/h range
Strobe Light Flash Rate115/min.
Strobe Light Operating Life200 hr.
Transmit Power Output100 mW
VoltagePower Supply: 9 VDC; Strobe Light: 6 - 14 VDC; Sound Alarm: 5 - 15 VDC
Weight (English)1.75 lb.
Weight (Metric)0.79 kg
Product TypeArea Radiation Monitoring System for Nuclear Power Plants
Radiation TypeGamma
Unit SizeEach