Beveled Non-Filtered Pipette Tips, 1-200 μL, QSP 200
Actual product may vary

Beveled Non-Filtered Pipette Tips, 1-200 μL, QSP 200

Filter and non-filter pipette tips in a complete portfolio for your evolving liquid transfer needs.
Have Questions?
Catalog NumberColorSterilityFormat
T070-QNaturalNon-sterileBagged
Catalog number T070-Q
HSN 39233090
Price (INR)
26,129.00
CS
Color:
Natural
Sterility:
Non-sterile
Format:
Bagged
Price (INR)
26,129.00
CS
Thermo Scientific™ QSP filter and non-filtered pipette tips are made from 100% virgin polypropylene utilizing state of the art molds and molding methods. Pipette tips are molded with high clarity resin and many include graduation marks and micro point design. Packaging options range from bulk to rack and rack sterile and include environmentally friendly options like tip reload systems and stack racks.

Featuring:

  • Micro point design tip orifice
  • High clarity resin
  • Graduation marks
  • Universal fit
  • Small micron filter
  • Modern and robust tip rack design

Clean Claims:

QSP Pipette Tips are free of:

  • Non-Sterile:
    • RNase/DNase
    • DNA
    • Pyrogen
  • E-Beam Sterilized:
    • RNase/DNase
    • DNA
    • Pyrogen/Endotoxin
    • Bioburden
    • PCR inhibitors
    • Endotoxin ATP

To order a clean claim lot-specific certification, contact your local dealer with the Cat. No and Lot No.

Recommend using with Finnpipette Digital pipette, but also compatible with other brands of pipettes.

Specifications
DescriptionQSP 200, Non-Filtered
Volume (Metric)200 μL
FilteredNon-filtered
SterilityNon-sterile
ColorNatural
Length (Metric)5.1 cm
Clean ClaimsDNase, RNase, DNA, and Pyrogen Free
FormatBagged
Tip StyleBeveled
Green FeaturesACT Ecolabel (Silver)
For Use With (Equipment)Universal Fit
GraduatedYes
Product LineQSP
MaterialVirgin Polypropylene
Unit SizeCS