Search
| Catalog Number | Packaging |
|---|---|
| T070RN-QPK also known as T070RN-Q | 960/Pk. |
Thermo Scientific™ QSP™ Soft Collar natural color Pipette Tip 200μl, packaged in rack non-sterile with clean claims; Rnase/Dnase, DNA, pyrogen, and endotoxin free. Universal fit on most pipettes.