QSP Filter and Non-Filtered Pipette Tips
Thermo Scientific™

QSP Filter and Non-Filtered Pipette Tips

Filter and non-filter pipette tips in a complete portfolio for your evolving liquid transfer needs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionVolume (Metric)Tip StyleSterility
T102RS-QPK
also known as T102RS-Q
QSP 10XL, Non-Filtered10 μLExtended LengthSterile
095-QPK
also known as 095-Q
QSP 5000, Non-Filtered5000 μLMacroNon-sterile
097-QPK
also known as 097-Q
QSP 10ML, Non-Filtered10 mLMacroNon-sterile
090-QPK
also known as 090-Q
QSP 5000, Non-Filtered5000 μLMacroNon-sterile
104-QPK
also known as 104-Q
QSP 10, Non-Filtered10 μLMicroPointNon-sterile
106-QPK
also known as 106-Q
QSP 300, Non-Filtered300 μLMicroPointNon-sterile
102-QPK
also known as 102-Q
QSP 10XL, Non-Filtered10 μLExtended LengthNon-sterile
110-NXL-QPK
also known as 110-NXL-Q
QSP 200XL, Non-Filtered200 μLExtended LengthNon-sterile
111-QPK
also known as 111-Q
QSP 1000, Non-Filtered1000 μLMicroPointNon-sterile
112NXL-QPK
also known as 112NXL-Q
QSP 1000, Non-Filtered1250 μLExtended LengthNon-sterile
111-N-QPK
also known as 111-N-Q
QSP 1000, Non-Filtered1000 μLMicroPointNon-sterile
112XL-QPK
also known as 112XL-Q
QSP 1000, Non-Filtered1000 μLExtended LengthNon-sterile
T070RNS-QPK
also known as T070RNS-Q
QSP 200, Non-Filtered200 μLBeveledSterile
T070RLN-QPK
also known as T070RLN-Q
QSP 200, Non-Filtered1 to 200 μLBeveledNon-sterile
T090RL-QPK
also known as T090RL-Q
QSP 200, Non-Filtered200 μLBeveledNon-sterile
T102R-QPK
also known as T102R-Q
QSP 10XL, Non-Filtered0.1 to 10 μLExtended LengthNon-sterile
T090RS-QPK
also known as T090RS-Q
QSP 200, Non-Filtered200 μLBeveledSterile
T090R-QPK
also known as T090R-Q
QSP 200, Non-Filtered200 μLBeveledNon-sterile
T104RL-QPK
also known as T104RL-Q
QSP 10, Non-Filtered10 μLMicroPointNon-sterile
T104RLS-QPK
also known as T104RLS-Q
QSP 10, Non-Filtered0.1 to 10 μLMicroPointSterile
T104R-QPK
also known as T104R-Q
QSP 10, Non-Filtered10 μLMicroPointNon-sterile
T104RS-QPK
also known as T104RS-Q
QSP 10, Non-Filtered10 μLMicroPointSterile
T106RL-QPK
also known as T106RL-Q
QSP 300, Non-Filtered300 μLMicroPointNon-sterile
T106RS-QPK
also known as T106RS-Q
QSP 300, Non-Filtered300 μLMicroPointSterile
T112NXLRLS-QPK
also known as T112NXLRLS-Q
QSP 1000, Non-Filtered1250 μLExtended LengthSterile
T112NXLR-QPK
also known as T112NXLR-Q
QSP 1000, Non-Filtered1250 μLExtended LengthNon-sterile
T112NXLRS-QPK
also known as T112NXLRS-Q
QSP 1000, Non-Filtered1250 μLExtended LengthSterile
T112XLRL-QPK
also known as T112XLRL-Q
QSP 1000, Non-Filtered1250 μLExtended LengthNon-sterile
T112XLRLS-QPK
also known as T112XLRLS-Q
QSP 1000, Non-Filtered1250 μLExtended LengthSterile
T112XLRS-QPK
also known as T112XLRS-Q
QSP 1000, Non-Filtered1250 μLExtended LengthSterile
T112XLR-QPK
also known as T112XLR-Q
QSP 1000, Non-Filtered1250 μLExtended LengthNon-sterile
TF102-10-QPK
also known as TF102-10-Q
QSP 10XL, Filtered10 μLExtended LengthSterile
TF140-200-QPK
also known as TF140-200-Q
QSP 200, Filtered200 μLMicroPointSterile
TF112-1000-QPK
also known as TF112-1000-Q
QSP 1000, Filtered100 to 1000 μLExtended LengthSterile
TF113-20-QPK
also known as TF113-20-Q
QSP 20, Filtered20 μLBeveledSterile
TF104-10-QPK
also known as TF104-10-Q
QSP 10, Filtered10 μLMicroPointSterile
TTW110RNS-QPK
also known as TTW110RNS-Q
QSP 200, Non-Filtered200 μLMicroPointSterile
TF113-100-QPK
also known as TF113-100-Q
QSP 100, Filtered100 μLBeveledSterile
TTW110RS-QPK
also known as TTW110RS-Q
QSP 200, Non-Filtered200 μLMicroPointSterile
TW110-N-QPK
also known as TW110-N-Q
QSP 200, Non-Filtered200 μLMicroPointNon-sterile
Catalog number T102RS-QPK
also known as T102RS-Q
Price (INR)
4,178.00
Pack of 960
Description:
QSP 10XL, Non-Filtered
Volume (Metric):
10 μL
Tip Style:
Extended Length
Sterility:
Sterile
Price (INR)
4,178.00
Pack of 960
Thermo Scientific™ QSP filter and non-filtered pipette tips are made from 100% virgin polypropylene utilizing state of the art molds and molding methods. Pipette tips are molded with high clarity resin and many include graduation marks and micro point design. Packaging options range from bulk to rack and rack sterile and include environmentally friendly options like tip reload systems and stack racks.

Featuring:

  • Micro point design tip orifice
  • High clarity resin
  • Graduation marks
  • Universal fit
  • Small micron filter
  • Modern and robust tip rack design

Clean Claims:
QSP Pipette Tips are free of:

  • Non-Sterile:
    • RNase/DNase
    • DNA
    • Pyrogen
  • E-Beam Sterilized:
    • RNase/DNase
    • DNA
    • Pyrogen/Endotoxin
    • Bioburden
    • PCR inhibitors
    • Endotoxin ATP

To order a clean claim lot-specific certification, contact your local dealer with the Cat. No and Lot No.

Recommend using with Finnpipette Digital pipette, but also compatible with other brands of pipettes.

Specifications
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor, and Pyrogen Free
DescriptionQSP 10XL, Non-Filtered
GraduatedYes
Length (Metric)4.6cm
ColorNatural
FilteredNon-filtered
For Use With (Equipment)Universal Fit
FormatHinged Rack
MaterialVirgin Polypropylene
Product LineQSP
SterilitySterile
Tip StyleExtended Length
Volume (Metric)10 μL
Unit SizePack of 960