Thermo Scientific™ QSP™ Multi-Channel Pipette Tip
Actual product may vary
Thermo Scientific™

Thermo Scientific™ QSP™ Multi-Channel Pipette Tip

Thermo Scientific™ QSP™ Multi-Channel Pipette Tips are designed for full volume multi-channel pipetting delivering even aspirations and dispenses.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilityFormat
T106RLS-QPK
also known as T106RLS-Q
SterileReload System
T106R-QPK
also known as T106R-Q
Non-sterileRack
Catalog number T106RLS-QPK
also known as T106RLS-Q
Price (INR)
-
Sterility:
Sterile
Format:
Reload System

Thermo Scientific™ QSP™ Multi-Channel natural color micropoint pipette Tip 300μl, graduated at 10, 50, 100, 200μl. Extended length 5.84cm. Packaged in reload system sterile with clean claims; Rnase/Dnase, DNA, pyrogen, endotoxin, ATP, PCR Inhibitor and bioburden free. Universal fit on most pipettes.

  • 300μl tip volume offers flexibiltiy in applications and pipetting techniques
  • Soft collar designed to work with multi-channel pipettes helps to seat pipette tips securely and evenly, offering uniform liquid aspiration and dispensing across all pipette channels
  • Universal fitting tips fit on a broad range of pipettes.
Specifications
Clean ClaimsSterile Electron Beam free of: Rnase/Dnase, DNA, pyrogen, endotoxin, ATP, PCR Inhibitor and bioburden
GraduatedYes
Graduations10, 50, 100, 200 μL
AutoclavableAutoclavable
ColorNatural
For Use With (Equipment)Universal Fit
FormatReload System
Material100% Virgin Polypropylene
Product LineQSP
SterilitySterile
Volume (Metric)300 μL
Unit SizePack of 960