Biocytin TMR (5-(and-6)-Tetramethylrhodamine Biocytin)
Invitrogen™
Biocytin TMR (5-(and-6)-Tetramethylrhodamine Biocytin)
Biocytin TMR (5-(and-6)-Tetramethylrhodamine Biocytin)
Have Questions?
Catalog NumberQuantity
T129215 mg
Catalog number T12921
Price (INR)
39,864.00
5 mg
Quantity:
5 mg
Price (INR)
39,864.00
5 mg
The water-soluble neuronal tracer 5-(and-6)-tetramethylrhodamine biocytin (biocytin TMR) can be detected in fixed tissues by using avidin or streptavidin probes. However, tetramethylrhodamine biocytin can also be observed directly in fixed or unfixed samples because the attached tetramethylrhodamine dye produces bright, orange-fluorescence (absorption/emission maxima ∼554/581 nm).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeBiocytins
Quantity5 mg
Shipping ConditionRoom Temperature
ConjugateClassic Dyes
FormSolid
Product LineInvitrogen
Unit Size5 mg
Contents & Storage
Store at room temperature and protect from light.
Citations & References (1)
Citations & References
Abstract
Evaluation of biotin-dye conjugates for use in an HPLC assay to assess relative binding of biotin derivatives with avidin and streptavidin.
Authors:Wilbur DS, Pathare PM, Hamlin DK, Frownfelter MB, Kegley BB, Leung WY, Gee KR
Journal:Bioconjug Chem
PubMed ID:10898581
In this investigation, studies were conducted to determine if size exclusion HPLC could be used to assess relative association rates (on-rates) and dissociation rates (off-rates) of biotin derivatives from avidin (Av) and streptavidin (SAv). For easy detection and quantification of biotin derivatives, molecules that can be detected by UV absorbance ... More