CSPD™ Substrate (25 mM Concentrate)
Invitrogen™
CSPD™ Substrate (25 mM Concentrate)
This concentrated CSPD™ substrate is a chemiluminescent alkaline phosphatase substrate for protein or nucleic acid blotting on nitrocellulose membranes. ThisRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
T204425 mL
T20981 mL
Catalog number T2044
Price (INR)
583,464.00
Each
Quantity:
25 mL
Price (INR)
583,464.00
Each
This concentrated CSPD™ substrate is a chemiluminescent alkaline phosphatase substrate for protein or nucleic acid blotting on nitrocellulose membranes. This substrate can also be used in solution-based assays.
This CSPD™ chemiluminescent substrate for alkaline phosphatase let you detect alkaline phosphatase and alkaline phosphatase-labeled molecules with unparalleled sensitivity, speed, and ease.
• Chemiluminescent substrates provide highly sensitive replacements for the widely used fluorogenic substrate methylumbelliferyl phosphate (MUP), and the colorimetric substrate p-nitrophenyl phosphate (pNPP).
• Low background luminescence coupled with high-intensity light output enables detection of alkaline phosphatase labels with the highest possible sensitivity and signal-to-noise.
This versatile chemiluminescent substrate exhibits high sensitivity in membrane-based applications such as Southern, Northern, and Western blotting. This substrate can also be used in solution-based assays such as immunoassays, DNA probe assays, enzyme assays, and reporter gene assays. Maximum light levels are reached in approximately 10 minutes and glow emission persists for several hours. Solution assays require chemiluminescent enhancers. Membrane-based assays may also require them for increased light output and sensitivity.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Southern, Northern, and Western Blotting
FormatBottle(s)
Product LineCSPD, NovaBright
Quantity25 mL
Shipping ConditionRoom Temperature
SubstrateAlkaline Phosphatase
FormLiquid
Substrate TypeAP (Alkaline Phosphatase) Substrate
Unit SizeEach
Contents & Storage
Store the product at 4°C.