Transferrin From Human Serum, Alexa Fluor™ Conjugate
Invitrogen™

Transferrin From Human Serum, Alexa Fluor™ Conjugate

Transferrin is a monomeric serum glycoprotein that binds to a specific receptor on the surface of vertebrate cells and delivers up to two Fe3+ atoms via receptor-mediated endocytosis—our labeled LDL complexes are useful tools for studying this phenomenon.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDye TypeQuantity
T23362Alexa Fluor Dyes5 mg
T13342Alexa Fluor Dyes5 mg
T2871Classic Dyes5 mg
T23364Alexa Fluor Dyes5 mg
T35352Alexa Fluor Dyes5 mg
T23365Alexa Fluor Dyes5 mg
T13343Alexa Fluor Dyes5 mg
T23366Alexa Fluor Dyes5 mg
Catalog number T23362
Price (INR)
38,354.00
Online offer
Ends: 31-Dec-2026
56,820.00
Save 18,466.00 (32%)
5 mg
Dye Type:
Alexa Fluor Dyes
Quantity:
5 mg
Price (INR)
38,354.00
Online offer
Ends: 31-Dec-2026
56,820.00
Save 18,466.00 (32%)
5 mg

Fluorescent transferrin for receptor-mediated endocytosis and recycling

Fluorescent transferrin conjugates are widely used to study transferrin receptor-mediated endocytosis, endosomal trafficking, and recycling using live-cell imaging or fixed-cell endpoint analysis. Transferrin binds transferrin receptor 1 (TfR1) at the cell surface and is internalized through clathrin-mediated endocytosis. Following endosomal acidification and iron release, apo-transferrin remains bound to TfR1 and the complex recycles to the plasma membrane, where apo-transferrin dissociates.

Why use fluorescent transferrin for endocytosis studies?

  • Track receptor-mediated endocytosis: Follow transferrin binding, TfR-mediated internalization, and intracellular trafficking.
  • Monitor endocytic recycling: Use fluorescent transferrin as a well-established marker of the endosomal recycling pathway.
  • Study trafficking kinetics: Follow uptake and recycling using live-cell imaging, pulse-chase experiments, or fixed-cell endpoint analysis.
  • Assess endosomal trafficking: Investigate trafficking events associated with endosomal acidification, cargo sorting, and membrane transport.
  • Compare trafficking pathways: Combine fluorescent transferrin with fluorescent LDL or other cargo markers to compare recycling and lysosomally directed pathways.
  • Enable multiplex fluorescence imaging: Choose from fluorescein and Alexa Fluor™ 488, 546, 555, 568, 594, 633, and 647 conjugates for multicolor imaging workflows.

Fluorescent transferrin enables a direct fluorescence readout of a well-characterized endocytic pathway, making it a versatile probe for studying receptor internalization, endosomal trafficking, and recycling.

Choose a fluorescent transferrin conjugate for your imaging workflow

Select from fluorescein and Alexa Fluor™ 488, 546, 555, 568, 594, 633, and 647 transferrin conjugates, with fluorescence spanning green to far-red wavelengths.

Multiple fluorophore options

  • Match your imaging system: Select a conjugate compatible with available excitation sources, emission filters, and detectors.
  • Support multicolor imaging: Choose spectrally distinct labels for use with antibodies, organelle markers, and other fluorescent probes.
  • Optimize experimental design: Select the excitation and emission profile that best fits your available fluorescence channels.
  • Use one biological probe across workflows: Study transferrin receptor trafficking with fluorophore options suitable for a range of fluorescence microscopy applications.

Applications of fluorescent transferrin

Fluorescent transferrin is a well-established probe for studying transferrin receptor-mediated endocytosis and intracellular trafficking.

Use fluorescent transferrin to:

  • Visualize receptor-mediated internalization and recycling using live-cell imaging or fixed-cell endpoint analysis
  • Track endosomal trafficking using fluorescence and confocal microscopy
  • Study trafficking events associated with endosomal acidification and changes in endosomal function
  • Investigate transferrin receptor dynamics, including appropriately designed FRET-based studies
  • Measure transferrin receptor binding in mammalian and parasite model systems
  • Compare recycling and lysosomal trafficking using complementary fluorescent cargo markers
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dye TypeAlexa Fluor Dyes
Excitation/Emission632/650
FormSolid
Protein FamilyTransferrin
Quantity5 mg
Shipping ConditionRoom Temperature
Product LineAlexa Fluor
Product TypeTransferrin
pH7.2
Unit Size5 mg
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Citations & References (19)

Citations & References
Abstract
CLIPR-59, a new trans-Golgi/TGN cytoplasmic linker protein belonging to the CLIP-170 family.
Authors:Perez F, Pernet-Gallay K, Nizak C, Goodson HV, Kreis TE, Goud B
Journal:J Cell Biol
PubMed ID:11854307
'The microtubule cytoskeleton plays a fundamental role in cell organization and membrane traffic in higher eukaryotes. It is well established that molecular motors are involved in membrane-microtubule interactions, but it has also been proposed that nonmotor microtubule-binding (MTB) proteins known as CLIPs (cytoplasmic linker proteins) have basic roles in these ... More
Geldanamycin stimulates internalization of ErbB2 in a proteasome-dependent way.
Authors:Lerdrup M, Hommelgaard AM, Grandal M, van Deurs B
Journal:J Cell Sci
PubMed ID:16352662
'The potent oncoprotein and receptor tyrosine kinase ErbB2 is remarkable because it resists efficient downregulation. However, ErbB2 can be downregulated by the HSP-90 inhibitor geldanamycin, but the underlying cellular mechanisms are uncertain. Apparently, delivery of ErbB2 to lysosomes, cleavage of the ErbB2 kinase domain and proteasomal activity are all processes ... More
Evaluation of disulfide reduction during receptor-mediated endocytosis by using FRET imaging.
Authors:Yang J, Chen H, Vlahov IR, Cheng JX, Low PS
Journal:Proc Natl Acad Sci U S A
PubMed ID:16950881
'Despite functional evidence for disulfide bond-reducing activity in endosomal compartments, the mechanistic details pertaining to such process (e.g., kinetics and sites of disulfide reduction) remain largely controversial. To address these questions directly, we have synthesized a previously uncharacterized fluorescent folate conjugate, folate-(BODIPY FL)-SS-rhodamine (folate-FRET), that changes fluorescence from red to ... More
Self-assembled, aptamer-tethered DNA nanotrains for targeted transport of molecular drugs in cancer theranostics.
Authors:Zhu G, Zheng J, Song E, Donovan M, Zhang K, Liu C, Tan W,
Journal:Proc Natl Acad Sci U S A
PubMed ID:23630258
'Nanotechnology has allowed the construction of various nanostructures for applications, including biomedicine. However, a simple target-specific, economical, and biocompatible drug delivery platform with high maximum tolerated doses is still in demand. Here, we report aptamer-tethered DNA nanotrains (aptNTrs) as carriers for targeted drug transport in cancer therapy. Long aptNTrs were ... More
Pincher, a pinocytic chaperone for nerve growth factor/TrkA signaling endosomes.
Authors:Shao Y, Akmentin W, Toledo-Aral JJ, Rosenbaum J, Valdez G, Cabot JB, Hilbush BS, Halegoua S
Journal:J Cell Biol
PubMed ID:12011113
A central tenet of nerve growth factor (NGF) action that is poorly understood is its ability to mediate cytoplasmic signaling, through its receptor TrkA, that is initiated at the nerve terminal and conveyed to the soma. We identified an NGF-induced protein that we termed Pincher (pinocytic chaperone) that mediates endocytosis ... More