Product Image
Trek Diagnostic System

Middlebrook 7H9 Broth 10ml 10 pcs

Middlebrook 7H9 Broth 10ml 10 pcs for use with Mycobacterium tuberculosis Sensititre Plates
Have Questions?
Catalog NumberQuantity
T344110
Catalog number T3441
Price (INR)
-
Quantity:
10
VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on. For Research Use Only. Not for use in diagnostic procedures.
Specifications
Certifications/ComplianceARTG
For Use With (Application)Trek Sensititre
FormatBox
Media TypeMiddlebrook Broth
No. per Pack10
Packaging Type10 x 10 mL Tubes
Quantity10
Shelf Life1 Year
Manufacturing OriginUnited Kingdom
Product TypeMiddlebrook 7H9
SpeciesMycobacterium tuberculosis
Unit SizeEach
Contents & Storage
2°C to 8°C