Campylobacter Test Organism Set
Thermo Scientific™
Campylobacter Test Organism Set
Perform simple, accurate validation of the Thermo Scientific™ Sensititre™ Campylobacter MIC Plate with the Thermo Scientific Campylobacter Test Organism Set.
Have Questions?
Catalog number T5104
Price (INR)
-
Perform simple, accurate validation of the Thermo Scientific™ Sensititre™ Campylobacter MIC Plate with the Thermo Scientific Campylobacter Test Organism Set.
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
DescriptionCampylobacter Test Organism Set
Quantity10/Pk.
Product TypeValidation Panel
SpeciesCampylobacter
Test TypeMinimum Inhibitory Concentration (MIC)
Unit SizeEach