NuPAGE™ WedgeWell™ Tris-Acetate Welcome Pack, 3 to 8%
Invitrogen™

NuPAGE™ WedgeWell™ Tris-Acetate Welcome Pack, 3 to 8%

The NuPAGE Tris-Acetate Welcome Pack, WedgeWell format provides all the necessary NuPAGE Tris-Acetate gels, buffers, and reagents you will need to begin using the Mini Gel Tank.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Wells
TA03815C15-well
TA03810A10-well
TA03812B12-well
Catalog number TA03815C
Price (INR)
48,282.00
Online offer
Ends: 31-Dec-2026
68,973.00
Save 20,691.00 (30%)
1 kit
No. of Wells:
15-well
Price (INR)
48,282.00
Online offer
Ends: 31-Dec-2026
68,973.00
Save 20,691.00 (30%)
1 kit
The NuPAGE Tris-Acetate Welcome Pack, WedgeWell format provides all the necessary NuPAGE Tris-Acetate gels, buffers, and reagents you will need to begin using the Mini Gel Tank.

The NuPAGE Tris-Acetate WedgeWell Welcome Pack provides all the necessary NuPAGE Tris-Acetate gels, buffers, and reagents you will need to begin using the Mini Gel Tank.

Welcome Pack contents

  • Mini Gel Tank (A25977)
  • NuPAGE Tris-Acetate WedgeWell mini gels (2 boxes, 20 gels)
  • NuPAGE Tris-Acetate SDS Running Buffer, 20X (LA0041)
  • NuPAGE LDS Sample Buffer, 4X (NP0007)
  • NuPAGE Sample Reducing Agent, 10X (NP0009)
  • HiMark Pre-stained Protein Standard (LC5699)

About the Mini Gel Tank

The Mini Gel Tank is compatible with all Novex, NuPAGE, and Bolt mini gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient, side-by-side gel loading and enhanced viewing during use. Run times may vary depending on buffer conditions and power supplies used.

About NuPAGE Tris-Acetate gels

NuPAGE Tris-Acetate protein gels provide excellent separation of large molecular weight proteins. NuPAGE Tris-Acetate gels are high performance polyacrylamide gels that simulate the denaturing or the native conditions of the traditional Laemmli system. The unique buffer formulation maintains a low operating pH during electrophoresis, resulting in superior resolution of proteins compared to traditional Tris-glycine SDS-PAGE gels.

For transferring your proteins to a membrane, we recommend using for NuPAGE Transfer buffer (NP0006) for traditional wet transfer using the Mini Blot Module (B1000). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 3 Gel Transfer Device (IB31001).

Convenient to use

NuPAGE Tris-Acetate WedgeWell-format gels have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0-mm-thick gels. The wedge-shaped wells also have larger openings, making sample loading easier.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Gel Percentage3 to 8%
Gel TypeTris-Acetate
No. of Wells15-well
Quantity1 Welcome Pk. kit
Separation Range40 to 500 kDa
Shipping ConditionApproved for shipment at room temperature and wet ice
Thickness (Metric)1.0 mm
For Use With (Equipment)XCell SureLock Mini-Cell
Gel SizeMini
Product LineNuPAGE
Unit Size1 kit

Frequently asked questions (FAQs)

What are the recommended maximum sample loading volumes for the new WedgeWell format holds for the midi gels?

For the 12+2 well large gels, the maximum sample loading volume is 100 µL. For the 12+2 well small gels, the maximum sample loading volume is 90 µL. For 20 well gels, the maximum sample loading volume is 60 µL and for the 26 well gels, the maximum sample loading volume is 40 µL.

Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.

I used one of your protein standards for a western transfer and noticed that some of the lower-molecular weight protein bands passed through the membrane. How can I resolve this issue?

- Decrease voltage, current or length of transfer time
- Make sure that the methanol concentration in the transfer buffer is proper; use a methanol concentration of 10-20% methanol removes the SDS from SDS-protein complexes and improves the binding of protein to the membrane.
- Make sure that the SDS concentration (if added) in the transfer buffer is proper, don't use more than 0.02-0.04% SDS. Using too much SDS can prevent binding of proteins to the membrane.
- Check the pore size of the membrane and the size of the target protein. Proteins smaller than 10 kDa will easily pass through a 0.45 µm pore size membrane. If proteins smaller than 10 kDa are of interest, it would be better to use a 0.2 µm pore size membrane.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

I used one of your protein standards for a western transfer and noticed that some of the higher-molecular weight bands transferred very poorly to the membrane. Can you offer some tips?

- Increase voltage, current or length of time for transfer
- SDS in the gel and in the SDS-protein complexes promotes elution of the protein from the gels but inhibits binding of the protein to membranes. This inhibition is higher for nitrocellulose than for PVDF. For proteins that are difficult to elute from the gel such as large molecular weight proteins, a small amount of SDS may be added to the transfer buffer to improve transfer. We recommend pre-equilibrating the gel in 2X Transfer buffer (without methanol) containing 0.02-0.04% SDS for 10 minutes before assembling the sandwich and then transferring using 1X transfer buffer containing 10% methanol and 0.01%SDS.
- Methanol removes the SDS from SDS-protein complexes and improves the binding of protein to the membrane, but has some negative effects on the gel itself, leading to a decrease in transfer efficiency. It may cause a reduction in pore size, precipitation of some proteins, and some basic proteins to become positively charged or neutral. Make sure that the methanol concentration in the transfer buffer is not more than 10-20% and that high-quality, analytical grade methanol is used.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

I used one of your pre-stained standards on a Tris-Glycine gel and noticed that the molecular weights of the proteins were different than on a NuPAGE Bis-Tris gel. What is the reason for this?

Pre-stained standards have a dye that is covalently bound to each protein that will result in the standard migrating differently in different buffer systems (i.e., different gels). As a result, using a pre-stained standard for molecular weight estimation will only give the apparent molecular weight of the protein. Pre-stained standards may be used for molecular weight approximation, confirming gel migration and estimating blotting efficiency but for accurate molecular weight estimation, an unstained standard should be used.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

I used one of your protein standards and am seeing some extra bands in the lane. Can you offer some suggestions?

- While loading, take care to make sure that there is no cross-contamination from adjacent sample lanes.
- Make sure that the correct amount of standard is loaded per lane. Loading too much protein can result in extra bands and this is a problem especially with silver-stained gels.
- Improper storage of the standard or repeated freeze/thawing can result in protein degradation.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.