NuPAGE™ WedgeWell™ Tris-Acetate XCell SureLock™ Mini-Cell Welcome Pack, 3 to 8%
Invitrogen™

NuPAGE™ WedgeWell™ Tris-Acetate XCell SureLock™ Mini-Cell Welcome Pack, 3 to 8%

The NuPAGE Tris-Acetate Welcome Pack, WedgeWell format provides all the necessary NuPAGE Tris-Acetate gels, buffers, and reagents you will need to begin using the XCell SureLock Mini-Cell.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Wells
TA03815F15-well
TA03810D10-well
TA03812E12-well
Catalog number TA03815F
Price (INR)
47,760.00
Online offer
Ends: 31-Mar-2026
79,599.00
Save 31,839.00 (40%)
1 kit
No. of Wells:
15-well
Price (INR)
47,760.00
Online offer
Ends: 31-Mar-2026
79,599.00
Save 31,839.00 (40%)
1 kit
The NuPAGE Tris-Acetate Welcome Pack, WedgeWell format provides all the necessary NuPAGE Tris-Acetate gels, buffers, and reagents you will need to begin using the XCell SureLock Mini-Cell.

The NuPAGE Tris-Acetate WedgeWell Welcome Pack provides all the necessary NuPAGE Tris-Acetate gels, buffers, and reagents you will need to begin using the XCell SureLock Mini-Cell.

Welcome Pack contents

  • XCell SureLock Mini-Cell (EI0001)
  • NuPAGE Tris-Acetate mini WedgeWell gels (2 boxes, 20 gels)
  • NuPAGE Tris-Acetate SDS Running Buffer, 20X (LA0041)
  • NuPAGE LDS Sample Buffer, 4X (NP0007)
  • NuPAGE Sample Reducing Agent, 10X (NP0009)
  • HiMark Pre-stained Protein Standard (LC5699)

About the XCell SureLock Mini-Cell

The XCell SureLock Mini-Cell is a vertical mini-protein gel electrophoresis system compatible with all Invitrogen mini precast gels and SureCast handcast gels. The Mini-Cell is simple, sturdy, and easy to use. Each XCell SureLock Mini-Cell can accommodate up to two gels per run. Run times may vary depending on buffer conditions and power supplies used.

About NuPAGE Tris-Acetate gels

NuPAGE Tris-Acetate protein gels provide excellent separation of large molecular weight proteins. NuPAGE Tris-Acetate gels are high performance polyacrylamide gels that simulate the denaturing or the native conditions of the traditional Laemmli system. The unique buffer formulation maintains a low operating pH during electrophoresis, resulting in superior resolution of proteins compared to traditional Tris-glycine SDS-PAGE gels.

For transferring your proteins to a membrane, we recommend using for NuPAGE Transfer buffer (NP0006) for traditional wet transfer using the XCell II Blot Module (EI9051). Alternatively, rapid semi-dry transfer can be performed using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 3 Gel Transfer Device (IB31001).

Convenient to use

NuPAGE Tris-Acetate WedgeWell-format gels have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0-mm-thick gels. The wedge-shaped wells also have larger openings, making sample loading easier.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Gel Percentage3 to 8%
Gel TypeTris-Acetate
No. of Wells15-well
Quantity1 Welcome Pk. kit
Separation Range40 to 500 kDa
Shipping ConditionApproved for shipment at room temperature and wet ice
Thickness (Metric)1.0 mm
For Use With (Equipment)XCell SureLock Mini-Cell
Gel SizeMini
Product LineNuPAGE
Unit Size1 kit