Product Image
Thermo Scientific™
Oxoid™ TST-RPLA Kit
Oxoid TST-RPLA Kit detects staphylococcal toxic-shock syndrome toxin (TST) by reversed passive latex agglutination.
Have Questions?
Catalog NumberQuantity
TD0940A20 Tests/Kit
Catalog number TD0940A
Price (INR)
-
Quantity:
20 Tests/Kit
Oxoid TST-RPLA Kit detects staphylococcal toxic-shock syndrome toxin (TST) by reversed passive latex agglutination.
Specifications
Quantity20 Tests/Kit
Specimen TypeStaphylococcal Toxic-Shock Syndrome Toxin (TST)
Product TypeMicrobiology Media
SpeciesStaphylococcus species
Test TypeReverse Passive Latex Agglutination
Unit SizeEach
Contents & Storage
2°C to 8°C