Product Image
Thermo Scientific™
Oxoid™ VTEC-RPLA Toxin Detection Kit
Uses reverse passive latex agglutination to detect verocytotoxin VT1 and VT2 produced by E. coli cultured from food and fecal specimens.
Have Questions?
Catalog NumberQuantity
TD0960A20 Tests/Kit
Catalog number TD0960A
Price (INR)
-
Quantity:
20 Tests/Kit
Thermo Scientific™ Oxoid VTEC-RPLA Toxin Detection Kit uses reverse passive latex agglutination to detect verocytotoxin VT1 and VT2 produced by E. coli cultured from food and fecal specimens.
Specifications
DescriptionVTEC-RPLA Toxin Detection Kit
Quantity20 Tests/Kit
Unit SizeEach
Contents & Storage
2°C to 8°C