Complete Outdoor Enclosure for the TEOM™ Series 1405
Protect Thermo Scientific™ TEOM™ Series 1405 Ambient Particulate Monitors from extreme climates with Thermo Scientific™ Complete Outdoor Enclosure for the TEOM Series 1405.
Have Questions?
Catalog Number
Description
TEOMSERIES1405ENCL
Complete Enclosure for TEOM Series 1405
Catalog number TEOMSERIES1405ENCL
Price (INR)
-
Description:
Complete Enclosure for TEOM Series 1405
Protect Thermo Scientific™ TEOM™ Series 1405 Ambient Particulate Monitors from extreme climates with the Thermo Scientific™ Complete Outdoor Enclosure for the TEOM Series 1405. Designed for use with the TEOM 1405 Series of analyzers, the Complete Outdoor Enclosure provides full weather protection.
The Thermo Scientific Complete Outdoor Enclosure for the TEOM Series 1405 monitors is designed to protect the instrument from extreme climates. Outfitted with a combination heating and air conditioning unit, the housing provides the proper weather-proof environment to ensure long-term performance.
Key Features:
Equipped with a combination heating and air conditioning unit
Full weather protection withstands extreme climates
Cost-effective alternative to permanent shelters
Flexible design will accommodate the TEOM Series 1405 with FDMS
Sample seal tube collar is available for the 1-1/4 in. flow splitter as well as the 1/2 in. diameter sample tubing
Cost-effective alternative to permanent shelters especially in locations where a fixed shelter is not available for installation
Corrosion-free aluminum composition and 2 in. thick insulation
Locking door latches, a door stop, vibration-free mounting for the sample pump available
Additional 18 in. and/or 10 in. shelves for additional hardware
Compatible with:
TEOM™ 1405 Monitor
TEOM™ 1405-D Monitor
TEOM™ 1405-DF Monitor
TEOM™ 1405F Monitor
Specifications
DescriptionComplete Enclosure for TEOM Series 1405
Outputs6-way outlet strip with power surge protector (Quantity 1)