QSP Low Retention Non-Filtered Pipette Tips
Actual product may vary

QSP Low Retention Non-Filtered Pipette Tips

Low retention pipette tips with a hydrophobic inner surface to reduce liquid retention and maintain pipetting precision.
Have Questions?
Catalog NumberVolume (Metric)Tip StyleFormatSterility
TLR106-Q300 μLMicroPointBulkNon-sterile
Catalog number TLR106-Q
HSN 39269099
Price (INR)
3,855.00
PK
Volume (Metric):
300 μL
Tip Style:
MicroPoint
Format:
Bulk
Sterility:
Non-sterile
Price (INR)
3,855.00
PK
Dispensing certain liquids can form a film on your tip, which can affect assay accuracy and precision. Thermo Scientific™ QSP low retention pipette tips offer a solution with a hydrophobic inner surface that reduces liquid retention.

Featuring:

  • Minimize the formation of films on inside of tip
  • Reduce sample retention
  • Improve accuracy and precision
  • Improve sample recovery
  • High clarity pipette tips
  • Variety of packaging options, including environmentally friendly tip reload systems

Clean Claims:

QSP Pipette Tips are free of:

  • Non-Sterile:
    • RNase/DNase
    • DNA
    • Pyrogen
  • E-Beam Sterilized:
    • RNase/DNase
    • DNA
    • Pyrogen/Endotoxin
    • Bioburden
    • PCR inhibitors
    • Endotoxin ATP

To order a clean claim lot-specific certification, contact your local dealer with the Cat. No and Lot No.

Recommend using with Finnpipette Digital pipette, but also compatible with other brands of pipettes.

Specifications
DescriptionQSP 300, Non-Filtered
Volume (Metric)300 μL
FilteredNon-filtered
SterilityNon-sterile
ColorNatural
Length (Metric)5.8cm
Clean ClaimsDNase, RNase, DNA, and Pyrogen Free
FormatBulk
Tip StyleMicroPoint
For Use With (Equipment)Universal Fit
GraduatedYes
Product LineQSP
Unit SizePK