See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
Home
▸
Shop All Products
▸
Western Blot Products
▸
Western Blot and Nucleic Acid Membranes and Filter Paper
▸
Owl™ Transfer Membranes
Actual product may vary
Thermo Scientific™
Owl™ Transfer Membranes
Perform Western blotting with hydrophobic Thermo Scientific™ Owl™ Transfer Membranes.
Have Questions?
Catalog Number
Packaging Type
TM
Transfer Membrane; Complete system
Catalog number
TM
Price (INR)
-
Request A Quote
Packaging Type:
Transfer Membrane; Complete system
Perform Western blotting with hydrophobic Thermo Scientific™ Owl™ Transfer Membranes.
Thermo Scientific offers Immobilon™-P, PVDF transfer membrane manufactured by Millipore™.
This membrane provides the highest signal-to-noise ratio, which allows the use of mild blocking procedures.
Immobilon-P has high protein retention, is compatible with all standard visualization methods and can be used for protein sequencing.
Immobilon is available in minigel and large-format gel sizes.
Specifications
Description
Transfer Membrane; Complete system
For Use With (Application)
Western blotting
Packaging Type
Transfer Membrane; Complete system
Material
Immobilon-P, PVDF
Product Line
Owl