Q Exactive Series eLearning Courses
Thermo Scientific™

Q Exactive Series eLearning Courses

The Q Exactive Series eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning courses which can cover different topics such as foundational theory and/or basic operations principle and/or user maintenance and troubleshooting.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCourse TopicCourse Level
TQEXFTHFoundational TheoryFoundation
TQEXBOPBasic OperationsRoutine
Catalog number TQEXFTH
Price (INR)
-
Course Topic:
Foundational Theory
Course Level:
Foundation

The Q Exactive Series eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument.

The Q Exactive Series User Maintenance and Troubleshooting eLearning course provides recommended maintenance guidelines and troubleshooting tips to maintain your Thermo Scientific™ Q Exactive™ Series Hybrid Quadrupole-Orbitrap™ Mass Spectrometer.

The Q Exactive Series Basic Operations eLearning course
Provides a novice user with instructions on how to operate the Q Exactive™ Series Hybrid Quadrupole-Orbitrap™ Mass Spectrometer.

Topics included in this course:

  • How to perform a system calibration with the BioPharma option, including guidelines on how to ensure spray stability, manipulating HESI source conditions, positive and negative mode calibrations, custom mass accuracy calibration, and high mass range (HMR) calibrations.
  • How to perform a sample injection using a standard configuration of a Thermo Scientific™ Vanquish™ Flex LC system in connection with a Q Exactive™ Plus Hybrid Quadrupole-Orbitrap™ MS.

The Q Exactive™ Series Hybrid Quadrupole-Orbitrap™ Mass Spectrometer Foundational Theory eLearning course
Visits each of the major components within the Q Exactive instruments to answer two questions: what does it do and how does it work?

  • The components include the ionization source, quadrupole, C-trap, HCD cell, Orbitrap, and more.
  • Discussions are technical enough to describe the major functions and principles without diving deep into the underlying physics and electronics.
  • This module targets users with some familiarity with mass spectrometry and chemistry concepts.
Specifications
Access Availablity30 days upon end user first access to the course
Course TopicFoundational Theory
FormatSelf-paced eLearning
LanguagesEnglish
Course LevelFoundation
Duration60 to 120 min.
For Use With (Equipment)Q Exactive
Product LineThermo Scientific
Product TypeMass Spectrometry
Target AudienceIntermediate user seeking to benefit from good instrument performance
Training LocationDigital access through Online Learning Management System
Unit SizeEach