Determine quantity of alkaline phosphatase in serum or plasma.
Have Questions?
Catalog Number
Quantity
TR11320
1 x 125 mL R1, 1 x 35 mL R2
Catalog number TR11320
Price (INR)
-
Quantity:
1 x 125 mL R1, 1 x 35 mL R2
Determine quantity of alkaline phosphatase in serum or plasma in vitro with Thermo Scientific™ Alkaline Phosphatase Reagents.
Features:
Utilizes the Tris/Carb buffer system
Reagent is water reconstituted with 30 days reconstituted stability
Variety of kit sizes make it suitable for all laboratory needs
Suitable for manual use or on automated clinical chemistry analyzers
Applications for a wide range of automated analyzers available on request
Notes: Elevations in serum activity of alkaline phosphatase commonly originate from the liver and bone and are of interest in the diagnosis of hepatobiliary disorders and bone disease associated with increased osteoblastic activity.