Product Image
Thermo Scientific™

Total Cholesterol Reagents

Determine quantity of cholesterol in serum in vitro with Thermo Scientific™ Total Cholesterol Reagents.
Have Questions?
Catalog NumberQuantity
TR134212 x 125 mL
Catalog number TR13421
Price (INR)
-
Quantity:
2 x 125 mL
Determine quantity of cholesterol in serum in vitro with Thermo Scientific™ Total Cholesterol Reagents.
Measurement of serum cholesterol levels can serve as an indicator of liver, biliary or thyroid function, intestinal absorption, adrenal disease and predisposition toward coronary artery disease. Increased levels are associated with nephrosis, diabetes mellitus and obstructive jaundice. When used in conjunction with triglyceride, HDL and LDL determinations, total cholesterol assays provide valuable information for the prediction of coronary artery disease. Decreased levels may be observed in cases of hyperthyroidism, certain anemias, malabsorption and wasting syndromes.
  • Up to 12 months reconstituted stability and various kit sizes to minimize reagent wastage
  • Suitable for manual use or on automated clinical chemistry analyzers
  • Applications for a wide range of automated analyzers available on request
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Certifications/ComplianceCE Marked
DescriptionInfinity Cholesterol 2 x 125mL
LinearityUp to 20mmol/L (774mg/dL)
Methods CompatibilityEndpoint
Quantity2 x 125 mL
Sample TypePlasma, Serum, Serum or Plasma
StabilityUntil expiry at 2-8°C
TypeReagent
CE MarkerYes
Detectable AnalytesCholesterol
Unit SizeEach