Product Image
Thermo Scientific™

Infinity Glucose Hexokinase 2 x 125mL

Determine quantity of glucose in human serum, plasma or urine in vitro with Thermo Scientific™ Glucose Hexokinase Reagents.
Have Questions?
Catalog number TR15421
Price (INR)
-
Determine quantity of glucose in human serum, plasma or urine in vitro with Thermo Scientific™ Glucose Hexokinase Reagents.

Diabetes mellitus is a group of metabolic disorders of carbohydrate metabolism in which glucose is underutilized, producing hyperglycemia. Measurement of blood glucose levels plays a key role in the diagnosis and management of the disease. Increased glucose levels may also be observed with hyperactivity of the thyroid, pituitary, or adrenal glands. Decreased levels are present in cases of insulin overdose, insulin secreting tumors, myxedema, hypopituitarism, hypoadrenalism, and conditions interfering with glucose absorption.

  • Up to 18 months of stability from the date of manufacture
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Certifications/ComplianceCE Marked
DescriptionInfinity Glucose Hexokinase
Linearity0 to 810mg/dL
Methods CompatibilityEndpoint
Quantity2 x 125 mL
StabilityStable Liquid
State at Room TemperatureLiquid
CE MarkerYes
Unit SizeEach
Contents & Storage
2°C to 8°C