Product Image
Thermo Scientific™

Amylase Reagent, Infinity Amylase EPS 2 x 125mL

The Amylase Reagent is suitable for manual use or on automated clinical chemistry analyzers.

Have Questions?
Catalog number TR25421
Price (INR)
-
Determine quantity of amylase in serum or urine in vitro with Thermo Scientific™ Amylase Reagent, suitable for manual use or on automated clinical chemistry analyzers.

Amylase is most frequently measured in the diagnosis of acute pancreatitis when serum levels may be grossly elevated. Hyperamylasemia is associated with, but not limited to, other acute abdominal disorders, biliary tract disease, diabetic ketoacidosis, severe glomerular dysfunction, salivary gland disorders, ruptured ectopic pregnancy and macroamylasemia. Applications for a wide range of automated analyzers are available on request.

For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Certifications/ComplianceCE Marked
DescriptionInfinity Amylase EPS
Linearity10 to 2000U/L
Methods CompatibilityKinetic
Quantity2 x 125 mL
StabilityStable Liquid
State at Room TemperatureLiquid
CE MarkerYes
Unit SizeEach
Contents & Storage
2°C to 8°C