Product Image
Thermo Scientific™

Creatinine Reagents

Determine the quantity of creatinine in human serum.
Have Questions?
Catalog NumberQuantity
TR351212 x 125 mL
Catalog number TR35121
Price (INR)
-
Quantity:
2 x 125 mL
Determine the quantity of creatinine in human serum in vitro with Thermo Scientific™ Creatinine Reagents.
Creatinine is a non-protein nitrogenous metabolite that is cleared from the body by the kidney and is used to assess renal function. Elevated levels are observed in acute nephritis, renal failure, urinary tract obstruction, shock, dehydration, congestive heart failure and diabetes. This reagent offers up to 18 months of stability from manufacture date, and applications are available for most automated chemistry analyzers.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Certifications/ComplianceCE Marked
DescriptionInfinity Creatinine (Modified Jaffe)
Linearity0 to 20mg/dL
Methods CompatibilityRate
Quantity2 x 125 mL
Sample TypeSerum
StabilityStable Liquid
State at Room TemperatureLiquid
TypeReagent
CE MarkerYes
Detectable AnalytesCreatinine
FormLiquid
Unit SizeEach
Contents & Storage
2°C to 8°C