Product Image
Thermo Scientific™

Lithium Reagent

Assure proper drug dosage is administered in the treatment of illnesses such as manic depression.
Have Questions?
Catalog NumberQuantity
TR660562 x 28 mL
Catalog number TR66056
Price (INR)
-
Quantity:
2 x 28 mL
Assure proper drug dosage is administered in the treatment of illnesses such as manic depression with Thermo Scientific™ Lithium Reagent.
  • Reagent is a liquid stable assay, offering 12 months stability.
  • Applications are available for most automated chemistry analyzers.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Certifications/ComplianceCE Marked
DescriptionInfinity Lithium
Linearity0.04 to 3mmol/L
Methods CompatibilityEndpoint
Quantity2 x 28 mL
Sample TypePlasma, Serum
StabilityStable Liquid
TypeReagent
CE MarkerYes
Detectable AnalytesLithium
FormLiquid
Unit SizeEach
Contents & Storage
2°C to 8°C