Product Image
Thermo Scientific™

Infinity™ ACE Assay

Measure angtiotensin-converting enzyme in serum with the Thermo Scientific™ Infinity™ ACE Assay.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
TR85101ACE Control Set
TR85056ACE (Angiotensin Converting Enzyme) Assay used to aid in the differential diagnosis of clinically active pulmonary sarcoidosis and for monitoring the effectiveness of steroid therapy
TR85201ACE Calibrator
Catalog number TR85101
Price (INR)
-
Description:
ACE Control Set
Measure angtiotensin-converting enzyme in serum with Thermo Scientific™ Infinity™ ACE Assay, a liquid stable reagent used to aid in the differential diagnosis of clinically active pulmonary sarcoidosis and for monitoring the effectiveness of steroid therapy.
  • Used for monitoring the effects of ACE inhibitors in the treatment of hypertension and heart failure
  • Applications available for most automated chemistry analyzers
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Certifications/ComplianceCE Marked
DescriptionACE Control Set
Linearity1 to 120U/L
Methods CompatibilityKinetic
Quantity6 x 1 mL
Sample TypeSerum
StabilityStable Liquid
State at Room TemperatureLiquid
TypeControl
CE MarkerYes
Detectable AnalytesACE (Angiotensin Converting Enzyme), Angtiotensin-converting Enzyme (ACE)
Unit SizeEach
Contents & Storage
2°C to 8°C