TruScan™ G3 Handheld Raman Analyzer
Thermo Scientific™

TruScan™ G3 Handheld Raman Analyzer

Rapid material identification testing for pharmaceutical and biotechnology manufacturing quality control/quality assurance processes.
Have Questions?
Catalog NumberSpectral Range
TRUSCANG3250 to 2875 cm-1
Catalog number TRUSCANG3
Price (INR)
-
Spectral Range:
250 to 2875 cm-1
The TruScan G3 Handheld Raman Analyzer continues the evolution of raw material ID testing for pharmaceutical and biotechnology manufacturing quality control processes requires speed, ease and efficiency to maintain continuous manufacturing. Samples are often brought to a quality control lab for testing, increasing costs and decreasing productivity. The TruScan G3 Handheld Raman Analyzer enables non-technical operators to rapidly perform raw material ID testing without the need of a lab, bringing lab-quality performance and results to any setting, whether that be a warehouse or loading dock.
  • Avoid compendial ID testing that requires extensive time and laboratory training by moving ID testing to the point of receipt.
  • Non-destructive, point and shoot sampling through plastic bags, glass containers, blister packs and clear gel caps.
  • Backwards compatible with Truscan RM Handheld Raman Analyzer validated methods, making deploying the TruScan G3 Handheld Raman Analyzer a quick method transfer for a near seamless experience.
  • A secured method for transferring of data via WIFI or wired connection for peace of mind.
  • No required, frequent hardware maintenance or consumables to operates
  • Enhanced 21 CFR Part 11 compliance
Specifications
Barcode Supported SymbologiesMost linear and 2D standards
Battery Type

Rechargeable internal lithium ion battery >3 hours operation

Certifications/ComplianceFDA 1040, 21 CFR part 11, CE certification, Ph. Eur. 8.7
Collection Optics

NA = 0.33, 18 mm working distance; 0.2 mm spot size

Connections

Ethernet, WiFi (optional)

DescriptionTruScan G3 Handheld Raman Analyzer
Dimensions (L x W x H)

7.8 x 4.6 x 1.8 in (19.9 x 11.8 x4.6 cm)

Exposure

Exposure time set automatically

Laser Excitation Wavelength785nm ± 0.5 (2 cm-1 line width, stability <0.1 cm-1)
Laser Output250 mW ±25
Operating Systems

OS: Windows 10 and 11; Browers: Edge and Chrome

Optional Accessories

Vial holder, extension probe, nose cone, hot swap batteries, hand strap, tablet holder

Ports

USBc and Power

Power SupplyDC Wall Adapter 100/240 V, 50/60 Hz
Spectral Range250 to 2875 cm-1
Spectral Resolution8 to 10.5 cm-1(FWHM) across range
Temperature Range (Metric)

-20°C to 50°C

Weight (English)

2.6 lb

Weight (Metric)

1.18 kg

Unit SizeEach