Reacti-Vap™ Evaporator PTFE-Coated Needles
Actual product may vary
Thermo Scientific
Reacti-Vap™ Evaporator PTFE-Coated Needles
Use these needles to deliver gas and evaporate samples on the Thermo Scientific™ Reacti-Vap™ Evaporator.
Have Questions?
Catalog NumberLength (Metric) Needle
TS-18784
also known as PIERCE #18784
102 mm
Catalog number TS-18784
also known as PIERCE #18784
HSN 90183210
Price (INR)
30,180.00
Each
Length (Metric) Needle:
102 mm
Price (INR)
30,180.00
Each
Use Thermo Scientific™ Reacti-Vap™ PTFE-Coated Needles to deliver gas and evaporate samples on Reacti-Vap Evaporators. These blunt-ended, 19-gauge, stainless steel needles reduce cross-contamination and corrosion when evaporating solvents that contain strong acids. The needles were specifically designed for Reacti-Vap Evaporators.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
MaterialPTFE-coated
Description9 Pack
Needle Gauge19 G
Length (English) Needle4 in.
Product TypeSyringe Needle
Length (Metric) Needle102 mm
For Use With (Equipment)Reacti-Vap Evaporators
Unit SizeEach