TSG Series Laboratory Freezers
TSG Series Laboratory Freezers
TSG Series Laboratory Freezers
TSG Series Laboratory Freezers
Thermo Scientific™

TSG Series Laboratory Freezers

TSG Auto-defrost Freezers offer tight temperature performance at -25°C setpoint with a 12-hour controller backup, remote alarm contacts and a 4-year parts and labor warranty. Available in single and double-door configurations, the freezers are sustainably made at a zero waste-to-landfill facility.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (English)Dimensions (DxWxH) ExteriorNo. of Doors
TSG5025FA50 cu.ft.29 x 53 x 58 in.2
TSG1225FA12 cu. ft.22 x 20 x 52 in.1
TSG2325FA23 cu ft.39 x 24 x 58 in.1
Catalog number TSG5025FA
Price (INR)
-
Capacity (English):
50 cu.ft.
Dimensions (DxWxH) Exterior:
29 x 53 x 58 in.
No. of Doors:
2
TSG Series Laboratory Freezers are built from the ground up to meet your cold storage needs, offering quiet operation, optimal performance, and sustainable design. The TSG Series is ready to go out of the box, complete with pre-installed casters, onboard alarms, a 12-hour controller backup system, and remote alarm contacts. These auto-defrost freezers also give you peace of mind knowing that your samples are secure with robust temperature performance, all backed by a 4-year parts and labor warranty.

Designed for sample integrity and security

  • Designed to safely store media, reagents, and other laboratory grade storage requiring tight temperature performance
  • Non-invasive adaptive defrost with time and temperature guided cycle
  • Easy to use microprocessor control with interior light, setpoint security and alarms
  • Standard door locks provide comfort and assurance

Enabling compliance

  • Tested and certified to conform with the design and safety standards of UL 61010, and CE Directive 2011/65/EU and 2015/863

Designed for an ergonomic and comfortable experience

  • Ergonomically designed door handle
  • Easy to clean and adjust epoxy coated shelves
  • Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
  • Self-closing door with 90° stop to assist with inventory loads

Verified sustainability

  • ACT Environmental Impact Factor Label Certified, providing third-party verification of the sustainable impacts of the product, its operations, and its end of life.
  • Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with EU F-GAS regulations, U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP), and other Global Environmental Regulations.
  • Manufactured in an award-winning, zero-waste to landfill facility

Ready for connectivity and monitoring

  • Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems

Make it your own

  • Extra and Alternate wire baskets, shelves, and drawers
  • E-Lock Adapter kits
  • 4-20 milliamp transmitters and other remote alarm modules

Services and support

  • Industry Leading 60-month compressor warranty (varies by country, please contact us for details)
  • Optional Validation Testing Programs include field-based temperature mapping and Installation Qualification (IQ) and Operation Qualification (OQ) programs
  • Optional Factory Acceptance Test results including QC Test Log Sheets and QC Temperature Graphs
Specifications
Access SecurityYes
Backup SystemsYes
Cabinet MaterialAluminum
Capacity (English) Shelf100 lb.
Capacity (Metric) Shelf45 kg
CastersYes
Chamber MaterialAluminum
DefrostAutomatic
Dimensions (DxWxH) Exterior29 x 53 x 58 in.
Door StyleDouble Solid
Electrical Requirements115 V/60 Hz
For Use With (Application)General Purpose
Frequency60 Hz
On-Board DataloggingYes
Plug TypeNEMA 5-15 P
RefrigerantR290
Shipping Weight (English)510 lb.
Shipping Weight (Metric)231 kg
Temperature Resolution1°C
Temperature SensorYes
Temperature (Setpoint)-25°C
Warranty4 Years (Parts and Labor)
Capacity (English)50 cu.ft.
Capacity (Metric)1416 L
Certifications/CompliancecULus, REACH, RoHS, EPA SNAP
CompressorBottom-mounted
Monitoring OptionsAlarm Module, Battery Back-up
No. of Doors2
Shelves8
StyleUpright
Temperature Range-15°C to -29°C
Voltage115 V
Unit SizeEach