Precision™ Shaking Water Baths
Thermo Scientific™

Precision™ Shaking Water Baths

Get shaking water baths that support a range of sensitive life science and QA/QC applications, from warming fragile reagents to tissue culture and genetic sequencing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Model
TSSWB15S15 LPrecision SWB 15S
TSSWB1515 LPrecision SWB 15
TSSWB2727 LPrecision SWB 27
TSDUB1515 LPrecision DUB 15
Catalog number TSSWB15S
Price (INR)
-
Capacity (Metric):
15 L
Model:
Precision SWB 15S

Benefit from Thermo Scientific™ Precision™ shaking water baths, which support a range of sensitive life science and QA/QC applications – from warming fragile reagents to tissue culturing and genetics sequencing. Easily clean and maintain your bath with new coil-free interior. The Precision shallow form shaking bath has a removable tray that allows for use with smaller sample containers. Precision Dubnoff shaking water baths are designed specifically for applications that require your samples to be incubated in a controlled atmosphere

  • Easily clean and maintain bath with coil-free design
  • Optimize scheduling with auto-on and auto-off timers
  • Help prevent bath overheating and damage with low-fluid protection
  • Conveniently save commonly used settings with four temperature and shaking speed presets
  • Help protect your work with audible alarms
  • Easily operate and monitor with icon-based graphical display
  • Accommodate taller labware with new hinged lid and extended height
  • Features next-generation shaker motor
  • Warranty varies by country, please contact us for details
Not for use for IVD/MD in some countries.
Specifications
Amperage9.0-10.5 / 4.3-5.0 (120V/230V)
Certifications/ComplianceUL, CE
Chamber Material304 Stainless Steel
Controller TypeDigital
Dimensions (L x W x H)15.5 x 24.9 x 9.8 in. (394 x 632 x 249 mm)
DisplayMonochrome LCD
Heating Capacity1200 w
IncludesStainless Steel Gable Cover and Rubber Duck
MaterialEpoxy Powder-coated Cold Rolled Steel
ModelPrecision SWB 15S
Temperature Presets4
Temperature Stability±0.1°C
Temperature Uniformity±0.05°C
Capacity (Metric)15 L
Electrical RequirementsGlobal Voltage 100/115 V 200/230 V 50/60 Hz
Temperature RangeAmbient +5° to 100°C
TypeShaking Water Bath
Unit SizeEach