TSX Series High-Performance Pharmacy Refrigerators
TSX Series High-Performance Pharmacy Refrigerators
TSX Series High-Performance Pharmacy Refrigerators
Thermo Scientific™

TSX Series High-Performance Pharmacy Refrigerators

Innovative refrigeration solutions for healthcare and research needs

Have Questions?
Change viewbuttonViewtableView
Catalog NumberDoor StylePlug TypeElectrical RequirementsDimensions (D x W x H) Interior
TSX2305PZGlassNEMA 6-15208 - 230 V 60 Hz28.5 x 24.0 x 58.0 in.
TSX2305PAGlassNEMA 5-15115V, 60Hz28.5 x 24 x 58 in.
TSX5005PADouble GlassNEMA 5-15115V, 60Hz28.5 x 52.5 x 58 in.
TSX5005PDDouble GlassNEMA 6-15208-230V, 60Hz28.5 x 52.5 x 58 in.
TSX5005PVDouble GlassCEE 7/7208-230V, 50Hz28.5 x 52.5 x 58 in.
TSX1205PVGlassCEE 7/7208-230V, 50Hz21.8 x 20.0 x 52.4 in.
TSX1205PYGlassNEMA 5-15115 V 60 Hz21.8 x 20.0 x 52.4 in.
TSX1205PZGlassNEMA 6-15208 - 230 V 60 Hz21.8 x 20.0 x 52.4 in.
TSX3005PVGlassCEE 7/7208-230V, 50Hz28.5 x 30.0 x 58.0 in.
TSX3005PYGlassNEMA 5-15115 V 60 Hz28.5 x 30.0 x 58.0 in.
TSX3005PZGlassNEMA 6-15208 - 230 V 60 Hz28.5 x 30.0 x 58.0 in.
TSX5005PYDouble GlassNEMA 5-15115 V 60 Hz28.5 x 52.5 x 58 in.
TSX2305PDGlassNEMA 6-15208-230V, 60Hz28.5 x 24 x 58 in.
TSX5005PZDouble GlassNEMA 6-15208 - 230 V 60 Hz28.5 x 52.5 x 58 in.
TSX2305PVGlassCEE 7/7208-230V, 50Hz28.5 x 24 x 58 in.
TSX2305PYGlassNEMA 5-15115 V 60 Hz28.5 x 24.0 x 58.0 in.
Catalog number TSX2305PZ
Price (INR)
-
Door Style:
Glass
Plug Type:
NEMA 6-15
Electrical Requirements:
208 - 230 V 60 Hz
Dimensions (D x W x H) Interior:
28.5 x 24.0 x 58.0 in.

Thermo Scientific™ TSX Series high-performance pharmacy refrigerators are designed with features that support sample protection and sustainability objectives for the storage of pharmaceuticals, vaccines, chemotherapy and other medical and pharmacy-grade storage requiring 2° to 8°C. The Series' V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.

Certified to NSF/ANSI 456 Vaccine Storage standard, the TSX series can help you meet regulations and guidelines from the CDC, World Health Organization, and other Health Ministries on vaccine storage and handling.

The TSX Series delivers the performance and quality you need, in addition to the low sound, low energy, and exceptional user experience you deserve.

The drive for sample integrity and security
  • Designed to safely store pharmaceuticals, vaccines, chemotherapy and other medical-grade storage requiring a temperature range of 2°C to 8°C
  • Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
  • Easy to use microprocessor control with interior light, setpoint security and alarms
  • Standard door locks provide comfort and assurance
Enabling compliance
  • Tested and Certified to NSF/ANSI 456 Vaccine Storage standard and supports the US CDC, WHO, and other global vaccine storage guidelines and recommendations
  • Tested to GMP Clean Room Class A / ISO 5 & 6 (ISO EN 14644-1) with easy to clean surfaces support USP <797> and <800> hazardous drug compounding and other clean room environments
  • Tested and certified to conform with the design and safety standards of UL 471, 61010, and CE Directive 2011/65/EU and 2015/863
Designed for an ergonomic and comfortable experience
  • Ergonomically designed door handle with a soft-touch grip
  • The microprocessor controller is angled to help prevent neck strain
  • Convenient, fully extendable and adjustable basket drawers
  • Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
  • Self-closing door with 90° stop to assist with inventory loads
Verified sustainability
  • ACT Environmental Impact Factor Label Certified, providing third-party verification of the sustainable impacts of the product, its operations, and its end of life.
  • Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with EU F-GAS regulations, U.S. Environmental Protection Agency’s Significant New Alternatives Policy (SNAP), and other Global Environmental Regulations.
  • Manufactured in an award-winning, zero-waste to landfill facility
Ready for connectivity and monitoring
  • Thermo Scientific™ DeviceLink™ Connect ready, enabling you to have a connected experience with your unit so you always—and anywhere— know the safety of your samples and the performance of your refrigerator or freezer.
  • Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems
Make it your own
  • Alternate door swing configuration and door types
  • Extra and alternate wire baskets, shelves, and drawers
  • Free-standing or factory-installed chart recorders
  • Stainless steel interior finish
  • E-Lock adapter kits
  • 4-20 milliamp transmitters and other remote alarm modules
  • Specialty, regional, and country specific plugs
Services and support
  • Industry Leading 120-month compressor warranty (varies by country, please contact us for details)
  • Included Certificate of Calibration to an NIST (National Institute of Standards and Technology, formerly National Bureau of Standards) traceable instrument includes a model number and serial number certificate
  • Optional Validation Testing Programs include in house and field-based temperature mapping and Installation Qualification (IQ) and Operation Qualification (OQ) programs
  • Optional Factory Acceptance Test results including QC Test Log Sheets and QC Temperature Graphs
*Data on file. Based on comparisons between TSX Series and previous generation Thermo Scientific models.
Specifications
Amperage15 A
Cabinet MaterialCold-rolled Steel
Chart RecorderOptional
DefrostAutomatic
Depth (English) Exterior37.8 in.
Depth (English) Interior28.5 in.
Depth (Metric) Exterior96 cm
Depth (Metric) Interior72.3 cm
Dimensions (D x W x H) Interior28.5 x 24.0 x 58.0 in.
Dimensions (L x W x H) Exterior37.8 x 28.0 x 78.5 in. (96.0 x 71.1 x 199.4 cm)
DisplayMicroprocessor
DrawersWire Basket Drawers
Electrical Requirements208 - 230 V 60 Hz
Exterior FinishPainted Steel
Frequency60 Hz
Height (English) Exterior78.5 in.
Height (English) Interior58 in.
Height (Metric) Exterior199.4 cm
Height (Metric) Interior147.3 cm
Monitoring OptionsAlarm, Recorder
No. of Drawers6
Product LineTSX Series
Rack Capacity6
RefrigerantR290
Shelves6
Shipping Weight (English)510 lb.
Shipping Weight (Metric)231 kg
Smart View CompatibilitySmart-Vue Compatible
TypePharmacy Refrigerator
Width (English) Exterior28 in.
Width (English) Interior24 in.
Width (Metric) Exterior61 cm
Width (Metric) Interior71.1 cm
Capacity (English)23 cu. ft.
Capacity (Metric)650 L
Certifications/ComplianceCE, cUL, UKCA
CompressorV-drive
Door StyleGlass
No. of Doors1
Plug TypeNEMA 6-15
StyleUpright
Temperature Range2°C to 8°C
Voltage208/230 V
Unit SizeEach