TSX Series High-Performance -20°C Manual Defrost Freezers
Actual product may vary
TSX Series High-Performance -20°C Manual Defrost Freezers
TSX Series High-Performance -20°C Manual Defrost Freezers
TSX Series High-Performance -20°C Manual Defrost Freezers
TSX Series High-Performance -20°C Manual Defrost Freezers
TSX Series High-Performance -20°C Manual Defrost Freezers
TSX Series High-Performance -20°C Manual Defrost Freezers
TSX Series High-Performance -20°C Manual Defrost Freezers
TSX Series High-Performance -20°C Manual Defrost Freezers
TSX Series High-Performance -20°C Manual Defrost Freezers
TSX Series High-Performance -20°C Manual Defrost Freezers
TSX Series High-Performance -20°C Manual Defrost Freezers
TSX Series High-Performance -20°C Manual Defrost Freezers
TSX Series High-Performance -20°C Manual Defrost Freezers
TSX Series High-Performance -20°C Manual Defrost Freezers
TSX Series High-Performance -20°C Manual Defrost Freezers
TSX Series High-Performance -20°C Manual Defrost Freezers
TSX Series High-Performance -20°C Manual Defrost Freezers
TSX Series High-Performance -20°C Manual Defrost Freezers
TSX Series High-Performance -20°C Manual Defrost Freezers
TSX Series High-Performance -20°C Manual Defrost Freezers
TSX Series High-Performance -20°C Manual Defrost Freezers
TSX Series High-Performance -20°C Manual Defrost Freezers
TSX Series High-Performance -20°C Manual Defrost Freezers
Thermo Scientific™

TSX Series High-Performance -20°C Manual Defrost Freezers

Thermo Scientific™ TSX Series high-performance -20°C manual defrost freezers offer a sustainable choice for clinical and laboratory storage
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPlug TypeElectrical RequirementsDimensions (D x W x H) Interior
TSX2320FZNEMA 6-15208 - 230 V 60 Hz28.5 x 24.0 x 58.0 in.
TSX2320FANEMA 5-15115V, 60Hz28.5 x 24 x 58 in.
TSX3020FVUK/Ireland or EU (CEE 7/7)208-230V, 50Hz28.5 x 30.0 x 58.0 in.
TSX3020FYNEMA 5-15115 V 60 Hz28.5 x 30.0 x 58.0 in.
TSX3020FZNEMA 6-15208 - 230 V 60 Hz28.5 x 30.0 x 58.0 in.
TSX2320FDNEMA 6-15208-230V, 60Hz28.5 x 24 x 58 in.
TSX2320FVCEE 7/7208-230V, 50Hz28.5 x 24 x 58 in.
TSX2320FYNEMA 5-15115 V 60 Hz28.5 x 24.0 x 58.0 in.
Catalog number TSX2320FZ
Price (INR)
-
Plug Type:
NEMA 6-15
Electrical Requirements:
208 - 230 V 60 Hz
Dimensions (D x W x H) Interior:
28.5 x 24.0 x 58.0 in.

Thermo Scientific™ TSX Series high-performance -20°C manual defrost freezers are designed are designed for applications in which even slight intermittent coil warming during auto defrost cannot be tolerated. The design supports sample protection and sustainability objectives for the storage of enzymes, reagents, pharmaceuticals, vaccines, biological and other commonly used laboratory and clinical materials. The Series' V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.

The TSX Series delivers the performance and quality you need, in addition to the low sound, low energy, and exceptional user experience you deserve.

The drive for sample integrity and security

  • Designed to safely store enzymes, reagents, pharmaceuticals, vaccines, biological and other commonly used laboratory and clinical materials requiring a tight temperature performance
  • Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
  • Cold wall convection cooling for uniform and stable temperatures
  • Easy to use microprocessor control with interior light, setpoint security and alarms
  • Standard door locks provide comfort and assurance

Enabling compliance

  • Tested to GMP Clean Room Class A / ISO 5 & 6 (ISO EN 14644-1) with easy to clean surfaces support USP <797> and <800> hazardous drug compounding and other clean room environments
  • Tested and certified to conform with the design and safety standards of UL 471, 61010, and CE Directive 2011/65/EU and 2015/863

Designed for an ergonomic and comfortable experience

  • Ergonomically designed door handle with a soft-touch grip
  • The controller is angled to help prevent neck strain
  • Automatic pressure equalization port the enables fast reentry
  • Easy to clean and adjust epoxy coated shelves
  • Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
  • Self-closing door with 90° stop to assist with inventory loads

Verified sustainability

  • US EPA ENERGY STAR Certified with up to 50% less energy usage than conventional refrigerant models*
  • ACT Environmental Impact Factor Label Certified, providing third-party verification of the sustainable impacts of the product, its operations, and its end of life.
  • Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with EU F-GAS regulations, U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP), and other Global Environmental Regulations.
  • Manufactured in an award-winning, zero-waste to landfill facility

Ready for connectivity and monitoring

  • Thermo Scientific™ DeviceLink™ Connect ready, enabling you to have a connected experience with your unit so you always—and anywhere— know the safety of your samples and the performance of your refrigerator or freezer.
  • Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems

Make it your own

  • Alternate Door swing configuration and door types
  • Extra and Alternate wire baskets, shelves, and drawers
  • Free-standing or factory-installed chart recorders
  • E-Lock Adapter kits
  • 4-20 milliamp transmitters and other remote alarm modules
  • Specialty, regional, and country specific plugs

Services and support

  • Industry Leading 120-month compressor warranty (varies by country, please contact us for details)
  • Optional Certificate of Calibration to an NIST (National Institute of Standards and Technology, formerly National Bureau of Standards) traceable instrument includes a model number and serial number certificate
  • Optional Validation Testing Programs include in house and field-based temperature mapping and Installation Qualification (IQ) and Operation Qualification (OQ) programs
  • Optional Factory Acceptance Test results including QC Test Log Sheets and QC Temperature Graphs

* Data on file. Based on comparisons between TSX Series and previous generation Thermo Scientific models.

*Data on file. Based on comparisons between TSX Series and previous generation Thermo Scientific models.
Specifications
Amperage15 A
Chart RecorderOptional
DefrostManual
Depth (English) Exterior38.9 in.
Depth (English) Interior28.5 in.
Depth (Metric) Exterior99 cm
Depth (Metric) Interior72.3 cm
Dimensions (D x W x H) Interior28.5 x 24.0 x 58.0 in.
Dimensions (L x W x H) Exterior38.9 x 28.0 x 78.5 in. (99.0 x 71.1 x 199.4 cm)
DisplayMicroprocessor
Door StyleSolid
Electrical Requirements208 - 230 V 60 Hz
Frequency60 Hz
Height (English) Exterior78.5 in.
Height (English) Interior58 in.
Height (Metric) Exterior199.4 cm
Height (Metric) Interior147.3 cm
Plug TypeNEMA 6-15
Product LineTSX Series
Rack Capacity4
RefrigerantR290
Shipping Weight (English)438 lb.
Shipping Weight (Metric)199 kg
Smart View CompatibilitySmart-Vue Compatible
TypeLaboratory Freezer
Width (English) Exterior71.1 in.
Width (English) Interior24 in.
Width (Metric) Interior61 cm
Capacity (English)23.3 cu. ft.
Capacity (Metric)659 L
Certifications/ComplianceCE, cUL
CompressorV-drive
Monitoring OptionsWireless
No. of Doors1
Shelves4
StyleUpright
Temperature Range-15°C to -25°C
Voltage208/230 V
Unit SizeEach